Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Violanthin | 40581-17-7 | 5 MG
SDP

Catalog No. 5000419803
Encompass_Preferred

Violanthin, isolated from the stems of *Dendrobium officinale*, exhibits potent antioxidant and antibacterial activities. It acts as an inhibitor of acetylcholinesterase (AChE) with an IC50 value of 79.80 μM. This compound is intended for research use only.

  • Molecular weight: 578.52
  • Formula: C27H30O14
  • Appearance: Solid
  • Color: White to yellow
  • Structure classification: Flavonoids, flavones, phenols, polyphenols
  • Initial source: Plants (Orchidaceae, Dendrobium officinale Kimura et Migo)
  • Exhibits potent antioxidant activity
  • Exhibits antibacterial activities
  • Inhibits acetylcholinesterase (AChE)

Catalog No. 50-004-19803 Supplier Medchemexpress LLC Supplier No. HYN68955MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Violanthin, isolated from the stems of *Dendrobium officinale*, exhibits potent antioxidant and antibacterial activities. It acts as an inhibitor of acetylcholinesterase (AChE) with an IC50 value of 79.80 μM. This compound is intended for research use only.

  • Molecular weight: 578.52
  • Formula: C27H30O14
  • Appearance: Solid
  • Color: White to yellow
  • Structure classification: Flavonoids, flavones, phenols, polyphenols
  • Initial source: Plants (Orchidaceae, Dendrobium officinale Kimura et Migo)
  • Exhibits potent antioxidant activity
  • Exhibits antibacterial activities
  • Inhibits acetylcholinesterase (AChE)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.