Learn More
Cytek Biosciences Inc VIOLET 450 MS CD4 (RM4-5) 100U
Supplier:  Cytek Biosciences Inc 750042U100
Encompass_Preferred
violetFluor 450 Anti-Mouse CD4 (RM4-5) Isotype: Rat IgG2a, kappa Apps: FC Size: 100 ug
Catalog No. 50-105-5101