Learn More
Biolegend Brilliant Violet 510™ anti-human CD279 (PD-1)
Supplier:  Biolegend 329932
Encompass_Preferred
Brilliant Violet 510™ anti-human CD279 (PD-1) [EH12.2H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests
Catalog No. NC0518712