Learn More
Biolegend Brilliant Violet 510™ anti-human CD11b
Supplier:  Biolegend 301334
Encompass_Preferred
Brilliant Violet 510™ anti-human CD11b [ICRF44]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: FC, ICC; Size: 100 tests
Catalog No. NC1592500