Learn More
Biolegend Brilliant Violet 785™ anti-human TCR α/β
Supplier:  Biolegend 306742
Encompass_Preferred
Brilliant Violet 785™ anti-human TCR α/β [IP26]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1888633