Learn More
Andwin Scientific VIOLET LABELING TAPE
SDP
Catalog No. 501126003
Encompass_Preferred
The 41.7' violet General-Purpose Laboratory Labeling Tape conveniently sticks to any surface and combats cool or hot temperature ranging from 23 to 121C (10 to 250F). This product can also be written using a pencil, pen, or marker.
Catalog No. 50-112-6003 Supplier Andwin Scientific Supplier No. 89098044
The 41.7' violet General-Purpose Laboratory Labeling Tape conveniently sticks to any surface and combats cool or hot temperature ranging from 23 to 121C (10 to 250F). This product can also be written using a pencil, pen, or marker.