Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Violet Red Bile Agar
SDP

Catalog No. 23001757 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-757 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-757 Supplier Hardy Diagnostics™ Supplier No. G78
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the detection and enumeration of coliforms in food, water, and dairy products

  • Violet Red Bile Agar is a selective medium used to detect and enumerate lactose-fermenting coliform microorganisms
  • The medium is recommended for use in the microbiological analysis of milk and other dairy products, and for use in the examination of water
  • The medium contains bile salts and crystal violet which serve as inhibitory agents toward some gram-positive microorganisms, especially staphylococc

Specifications

Appearance slightly opalescent
Certifications/Compliance ISO 13485
Color Reddish-purple
Content And Storage 2°C to 8°C
Detects Coliforms
Form Prepared Medium
Format Plate
Manufacturing Origin United States
Product Type Agar Plate
Target Organism Class Coliforms
Test Type Microbial Detection
Fill Volume 18 mL
Formulation Lactose - 10.0 g, Pancreatic Digest of Gelatin - 7.0 g, Sodium Chloride - 5.0 g, Yeast Extract - 3.0 g, Bile Salts No. 3 - 1.5 g, Neutral Red - 30.0 mg, Crystal Violet - 2.0 mg, Agar - 15.0 g
For Use With (Application) Microbiological Analysis, Coliform Detection
Incubation Conditions 35°C
Incubation Time 24 hr.
No. per Pack 10
Packaging Type Bag
Quantity 10 Plates
Dimensions (Dia. x H) 100 x 15 mm
Width (Metric) 100 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Enterobacter aerogenes, Escherichia coli
Protein Source Pancreatic Digest of Gelatin
Source Pancreatic Digest of Gelatin, Yeast
Manufacturing Quality ISO 13485
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.