Learn More
Bioworld Violet Red Bile Agar, 500 g
Catalog No. 50254391
Encompass_Preferred
Violet Red Bile AgarFor the enumeration of coliforms in food, milk, water and other sanitary materials Conforms to American Public Health Association (APHA) specificationspH: 7.4Working Concentration: 41.5 g/LDehydrated Appearance: red-beige, homogeneous powderPrepared Appearance: red-purple agarSoluble in boiling waterContains:Yeast Extract Pancreatic Digest of Gelatin Bile Salts #3 Lactose Sodium Chloride Neutral Red Crystal Violet Agar
Catalog No. 50-254-391 Supplier Bioworld Supplier No. 306200931
Violet Red Bile AgarFor the enumeration of coliforms in food, milk, water and other sanitary materials Conforms to American Public Health Association (APHA) specificationspH: 7.4Working Concentration: 41.5 g/LDehydrated Appearance: red-beige, homogeneous powderPrepared Appearance: red-purple agarSoluble in boiling waterContains:Yeast Extract Pancreatic Digest of Gelatin Bile Salts #3 Lactose Sodium Chloride Neutral Red Crystal Violet Agar