Learn More
MOLTOX Molecular Toxicology Inc Violet Red Bile Glucose Agar Plate, 20 mL/plate, 10 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948674
Encompass_Preferred
Violet Red Bile Agar (VRBA) is used for the enumeration of coliform organisms in dairy products. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-674 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S56
Violet Red Bile Agar (VRBA) is used for the enumeration of coliform organisms in dairy products. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.