Learn More
Alkali Scientific Violet Red Bile Glucose Agar 20mL Plate, 10/Pk
SDP
Catalog No. 5000663277
Encompass_Preferred
Violet Red Bile Glucose Agar 20mL Plate, 10/Pk. This selective medium is used for bacterial detection, particularly for the isolation of enteric organisms like coliforms and fecal streptococci. Ideal for laboratory settings requiring fast and accurate identification of bacteria, it is effective in the differentiation of various microbial species. Comes in a pack of 10 plates for convenient use in high-throughput environments.
Catalog No. 50-006-63277 Supplier Alkali Scientific Supplier No. VRBP00
Violet Red Bile Glucose Agar 20mL Plate, 10/Pk. This selective medium is used for bacterial detection, particularly for the isolation of enteric organisms like coliforms and fecal streptococci. Ideal for laboratory settings requiring fast and accurate identification of bacteria, it is effective in the differentiation of various microbial species. Comes in a pack of 10 plates for convenient use in high-throughput environments.