Learn More
Cytek Biosciences Inc VIOLET450 H/MCD45R(RA3-6B2)100
Supplier:  Cytek Biosciences Inc 750452U100
Encompass_Preferred
violetFluor 450 Anti-Human/Mouse CD45R (B220) (RA3-6B2) Isotype: Rat IgG2a, kappa Apps: FC Size: 100 ug
Catalog No. 50-105-5143