Learn More
Cytek Biosciences Inc Antibodies violetFluor™ 450 Anti-Mouse CD69 (H1.2F3) ….. Cat. No. 75-0691-U025 ….. Size 25 µg
Supplier:  Cytek Biosciences Inc 750691U025
Encompass_Preferred
Antibodies violetFluor™ 450 Anti-Mouse CD69 (H1.2F3) ….. Cat. No. 75-0691-U025 ….. Size 25 µgThe H1.2F3 antibody reacts with mouse CD69, a type II transmembrane glycoprotein also known as the Very Early Activation Antigen, EA-1, Leu23, Activation Inducer Molecule (AIM) and CLEC2C. CD69 is expressed as a 60 kDa disulfide-linked homodimer on activated T and B cells, NK cells, neutrophils and monocytes. Induction occurs rapidly upon activation. It is also constitutively expressed on platelets and a subset of thymocytes. CD69 acts as a co-stimulatory molecule involved in activation and proliferation of T cells, and may be a marker for thymocytes undergoing TCR-mediated positive selection.Co-stimulation with the H1.2F3 clone has been reported to enhance T cell and macrophage activation. Please choose the appropriate format for each application.
Catalog No. 50-210-3644