Learn More
AdipoGen Viomellein
SDP
Catalog No. 502561976
Encompass_Preferred
Chemical. CAS 55625-78-0. Formula C30H24O11. MW 560.5. Isolated from Aspergillus sulphureus. Genotoxic mycotoxin. Antibacterial Gram-positive and insecticidal. Aromatase inhibitor.
Catalog No. 50-256-1976 Supplier AdipoGen Supplier No. BVT0359C500
Chemical. CAS 55625-78-0. Formula C30H24O11. MW 560.5. Isolated from Aspergillus sulphureus. Genotoxic mycotoxin. Antibacterial Gram-positive and insecticidal. Aromatase inhibitor.