Learn More
Abcam Viomellein, Antibacterial agent, 500UG
Catalog No. 502705823
Encompass_Preferred
MW 560.5 Da, Purity >96%. Gram-positive antibacterial agent. Genotoxic mycotoxin. Dimeric quinone. Interacts with DNA. Shows bactericidal and insecticidal effects and induces mycotoxicosis in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5823 Supplier Abcam Supplier No. AB144383500UG
MW 560.5 Da, Purity >96%. Gram-positive antibacterial agent. Genotoxic mycotoxin. Dimeric quinone. Interacts with DNA. Shows bactericidal and insecticidal effects and induces mycotoxicosis in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement