Learn More
Waters Corp Vion™ IMS QTof PM Kit, Performance Maintenance, with Chemistry
Catalog No. 502391032 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit ensures optimum performance and trouble-free operation of the Vion IMS QTof system by facilitating annual maintenance tasks.
  • Inspection of fans
  • Replacement of fan filters (panels and electronics)
  • Cleaning of ion block and StepWave™ assemblies and replacement of consumables
  • Cleaning of entrance/exit plates of Cell 1, Cell 2, XS-ion optics, segmented ion guide, and DRE lens
  • Replacement of aperture plate
  • Emptying of fluidics and source waste bottles
  • Inspection of fluidics tubing and source waste line inside the instrument for leaks
  • Replacement of probe capillary
  • Servicing of ESI probe
  • Servicing of LockSpray probe and source housing
  • Updating of PCB firmware
  • Replacement of turbo oil cartridges (if required)
  • Replacement of TOF Penning gauge
  • Testing, cleaning, or replacement of the source door latch mechanism
Catalog No. 50-239-1032 Supplier Waters Corp Supplier No. 176004022
This kit ensures optimum performance and trouble-free operation of the Vion IMS QTof system by facilitating annual maintenance tasks.
  • Inspection of fans
  • Replacement of fan filters (panels and electronics)
  • Cleaning of ion block and StepWave™ assemblies and replacement of consumables
  • Cleaning of entrance/exit plates of Cell 1, Cell 2, XS-ion optics, segmented ion guide, and DRE lens
  • Replacement of aperture plate
  • Emptying of fluidics and source waste bottles
  • Inspection of fluidics tubing and source waste line inside the instrument for leaks
  • Replacement of probe capillary
  • Servicing of ESI probe
  • Servicing of LockSpray probe and source housing
  • Updating of PCB firmware
  • Replacement of turbo oil cartridges (if required)
  • Replacement of TOF Penning gauge
  • Testing, cleaning, or replacement of the source door latch mechanism