Learn More
Eurofins Discoverx VIP, 0.5 mg. (92-1074)
Supplier:  Eurofins Discoverx 921074
Encompass
Vasoactive intestinal peptide (VIP) is a 28 amino acid neuropeptide hormone that stimulates myocardial contractility, vasodilation and glycogenolysis, lowers arterial blood pressure, and relaxes the smooth muscles of the trachea, stomach, and gall bladder. VIP acts as an agonist for VIPR1 and VIPR2 (VPAC 1 and 2). Provided as 0.5 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9614