Learn More
Medchemexpress LLC HY-P1023 500ug Medchemexpress, VIP(6-28)(human, rat, porcine, bovine) CAS:69698-54-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1023500UG
Encompass_Preferred
Medchemexpress, HY-P1023 500ug VIP(6-28)(human, rat, porcine, bovine) CAS:69698-54-0 VIP(6-28)(human, rat, porcine, bovine) is an effective antagonist of the actions of exogenous vasoactive intestinal peptide ( VIP ) on cAMP. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4132