Learn More
Aviva Systems Biology Corp VIP Antibody - middle region (ARP45465_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45465_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VIP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human VIP. Alternate names: MGC13587, PHM27ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-53850