Learn More
Medchemexpress LLC VIP Human HEK293 5ug
SDP
Catalog No. 5000196732
Encompass_Preferred
Recombinant human vasoactive intestinal peptide (VIP) expressed in HEK293 cells and purified with a C-terminal 6xHis tag. This purified neuropeptide reagent is intended for research use in biochemical and cell-based studies that examine vasodilation, cardiac contractility, glycogenolysis, and smooth muscle physiology.
  • Expressed in mammalian (HEK293) cells for human-like post-translational processing.
  • Purified with a C-terminal 6xHis tag to facilitate detection and purification.
  • Provided in multiple microgram sizes for flexible experimental design.
  • Supplied with quality documentation (COA/datasheet) to support reproducibility.
  • Suitable for biochemical assays and cell-based functional studies.
Catalog No. 50-001-96732 Supplier Medchemexpress LLC Supplier No. HYP710615UG
Recombinant human vasoactive intestinal peptide (VIP) expressed in HEK293 cells and purified with a C-terminal 6xHis tag. This purified neuropeptide reagent is intended for research use in biochemical and cell-based studies that examine vasodilation, cardiac contractility, glycogenolysis, and smooth muscle physiology.
  • Expressed in mammalian (HEK293) cells for human-like post-translational processing.
  • Purified with a C-terminal 6xHis tag to facilitate detection and purification.
  • Provided in multiple microgram sizes for flexible experimental design.
  • Supplied with quality documentation (COA/datasheet) to support reproducibility.
  • Suitable for biochemical assays and cell-based functional studies.