Learn More
CPC Scientific H-His-Ser-Asp-Ala-Val-Phe-Thr-Asp-Asn-Tyr-Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2 0.5MG
SDP
Supplier:  CPC Scientific VIP001A
Encompass
A 28 amino acid peptide synthesized in the central nervous system and gastrointestinal tract. It belongs to the secretin glucagon-CRF family and is widely distributed in the central and peripheral nervous systems. In acting as both a neurotransmitter and hormone that has powerful hypotensive and vasodilatory effects, this peptide plays a wide role in a range of biological activities, including vaso- and bronchodilation, smooth muscle relaxation, and stimulation of secretin. When paired with the adrenegic drug phentolamine and injected, this peptide is expected to provide a new and effective solution for patients suffering from erectile dysfunction.SEQUENCE: H-His-Ser-Asp-Ala-Val-Phe-Thr-Asp-Asn-Tyr-Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2ONE-LETTER SEQUENCE: HSDAVFTDNYTRLRKQMAVKKYLNSILN-NH2MOLECULAR FORMULA: C147H238N44O42S1MOLECULAR WEIGHT: 3325.80STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [40077-57-4]SYNONYMS: Aviptadil, VIP (
Catalog No. 50-211-0607