Learn More
Aviva Systems Biology Corp VIPAR Antibody - middle region (ARP57586_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57586_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C14orf133. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human VIPAR. Alternate names: FLJ12707, SPE39, VIPAR, SPE-39, VPS16B, hSPE-39ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53851