Learn More
Thermo Scientific Viper SST 0.130 x 450 mm
Supplier:  Thermo Scientific 6040.2345
Encompass
Viper SST 0.130 x 450 mm, stainless steel, 1300 bar maximum pressure, for all HPLC and UHPLC systems
Catalog No. 50-134-7821