Learn More
Thermo Scientific Viper SST 0.180 x 350 mm
Supplier:  Thermo Scientific 6040.2375
Encompass
Viper™ Fingertight Fitting Systems, Stainless Steel, 0.18 mm x 350 mm, maximum pressure: 1300 bar
Catalog No. 50-134-7828