Learn More
Thermo Scientific Viper Fingertight Fitting Systems | Stainless Steel | 1 EA
Supplier:  Thermo Scientific 6040.2265
Encompass
Ensure optimal separations and maximize liquid chromatography LC performance with virtually zero-dead-volume connections, Easy to use and enable a fast tool-free fluidic setup of HPLC and UHPLC systems, Compatible with virtually any column and valve type, Suitable for system pressures up to 1310 bar (19000 psi)
Catalog No. 50-134-7800