Learn More
Thermo Scientific Viper Fingertight Fitting Systems | MP35N | 1 EA
Supplier:  Thermo Scientific 6042.2327
Encompass
Ferrule-free tightening design for virtually zero-dead-volume connections, Works with operating pressures up to 1517 bar, Suitable for temperatures up to 120C, Fingertight design, Flexible MP35N capillaries 1/32 in OD, No tools required, Compatible with valves and columns from virtually any manufacturer, Fits narrow connections such as 10-port valves, Available in different materials lengths and inner diameters
Catalog No. NC1515739