Learn More
Thermo Scientific Viper Fingertight Fitting Systems MP35N | | 1 EA
Catalog No. NC1515740 Shop All Thermo Scientific Products
Encompass
Robust LC Performance: Ferrule-free tightening design for virtually zero-dead-volume connections, Works with operating pressures up to 1517 bar, Suitable for temperatures up to 120C, Easy to Install: Fingertight design, Flexible MP35N capillaries 132 in OD, No tools required, Flexible: Compatible with valves and columns from virtually any manufacturer, Fits narrow connections such as 10-port valves, Available in different materials lengths and inner diameters, Recommended Use: Fluidic connections for analytical biocompatible LC systems, Thermo Scientific Vanquish Flex and Vanquish Horizon UHPLC systems and Thermo Scientifc UltiMate 3000 BioRS systems are equipped with the Viper Fingertight Fitting System with MP35N capillaries as standard
This item is not returnable. View return policy
Catalog No. NC1515740 Supplier Thermo Scientific Supplier No. 6042.2365
Robust LC Performance: Ferrule-free tightening design for virtually zero-dead-volume connections, Works with operating pressures up to 1517 bar, Suitable for temperatures up to 120C, Easy to Install: Fingertight design, Flexible MP35N capillaries 132 in OD, No tools required, Flexible: Compatible with valves and columns from virtually any manufacturer, Fits narrow connections such as 10-port valves, Available in different materials lengths and inner diameters, Recommended Use: Fluidic connections for analytical biocompatible LC systems, Thermo Scientific Vanquish Flex and Vanquish Horizon UHPLC systems and Thermo Scientifc UltiMate 3000 BioRS systems are equipped with the Viper Fingertight Fitting System with MP35N capillaries as standard