Learn More
Thermo Scientific Viper Fingertight Fitting Systems | MP35N | 1 EA
Catalog No. NC1515745 Shop All Thermo Scientific Products
Encompass
Ensure virtually zero-dead-volume connections for optimal liquid chromatography performance with easy-to-use biocompatible 180m I.D.fingertight fittings. Maximize liquid chromatography LC performance and ensure optimal separations with virtually zero-dead-volume connections. Thermo Scientific Viper Fingertight Fitting Systems with biocompatible MP35N capillaries are easy to use and enable a fast tool-free fluidic setup of biocompatible HPLC and UHPLC systems. These systems are compatible with virtually any column and valve type and suitable for fluidic connections with pressures up to 1517 bar 22000 psi.
This item is not returnable. View return policy
Catalog No. NC1515745 Supplier Thermo Scientific Supplier No. 6042.2375
Ensure virtually zero-dead-volume connections for optimal liquid chromatography performance with easy-to-use biocompatible 180m I.D.fingertight fittings. Maximize liquid chromatography LC performance and ensure optimal separations with virtually zero-dead-volume connections. Thermo Scientific Viper Fingertight Fitting Systems with biocompatible MP35N capillaries are easy to use and enable a fast tool-free fluidic setup of biocompatible HPLC and UHPLC systems. These systems are compatible with virtually any column and valve type and suitable for fluidic connections with pressures up to 1517 bar 22000 psi.