Learn More
Thermo Scientific Viper Capillary Column 0.13 x 350mm [1 EA]
Supplier:  Thermo Scientific 6040.2335
Encompass
0.13 x 350 MM stainless steel capillary gas chromatography column with polyimide external coating for analytical separation and high resolution of samples like food, environmental, petrochemical, and pharmaceutical with inertness, durability, flexibility, and protection.
Catalog No. NC1839321