Learn More
Thermo Scientific VIPER CPLLRY MP35N,180UM X 150
Catalog No. NC1429090 Shop All Thermo Scientific Products
Encompass
Viper Capillary MP35N, 180µm x 150mm
Catalog No. NC1429090 Supplier Thermo Scientific Supplier No. 6042.2315
Viper Capillary MP35N, 180µm x 150mm