Learn More
Thermo Scientific Viper PEEK 0.130 µm x 150 mm
Catalog No. 501347926 Shop All Thermo Scientific Products
Encompass
Viper PEEK tubing, for Fluidic connection for biocompatible HPLC and UHPLC systems, maximum pressure: 345 bar
Catalog No. 50-134-7926 Supplier Thermo Scientific Supplier No. 6041.5616
Viper PEEK tubing, for Fluidic connection for biocompatible HPLC and UHPLC systems, maximum pressure: 345 bar