Learn More
Thermo Scientific Viper Fingertight Fitting Systems MP35N | | 1 EA
Supplier:  Thermo Scientific 6042.2373
Encompass
- Ferrule-free tightening design for virtually zero-dead-volume connections- Operates at pressures up to 1517 bar- Suitable for temperatures up to 120C- Fingertight design- Flexible MP35N capillaries 1/32 in OD- No tools required- Compatible with virtually any valve and any column from any manufacturer- Fits narrow connections such as 10-port valves- Enables mixed use with different designs- Available in different materials, lengths, and inner diameters
Catalog No. NC1322458