Learn More
Thermo Scientific Viper Union [EA]
Supplier:  Thermo Scientific 6040.2304
Encompass
fluidic connection component for joining capillary tubing in liquid chromatography systems made of titanium with coned profile accepting tubing outer diameters 1/16IN to 1/8IN withstanding pressures up to 5,000PSI & temperatures 4°C to 150°C for leak-free connections between tubing with same or varying inner & outer diameters due to inert titanium construction suitable for wide range of solvents
Catalog No. 50-134-7805