Learn More
Thermo Scientific Dionex IC PEEK Viper Kits and Loops | N/A | 1 EA
Catalog No. NC1761297 Shop All Thermo Scientific Products
Encompass
Achieve consistent low dead volume connections; Ensure optimum chromatographic performance for ion chromatography (IC) applications; Conventional PEEK fittings can increase dead volume if the ferrule is incorrectly positioned or tubing poorly cut, resulting in reduced efficiencies and poor peak shapes; Provide worry-free finger-tight connections with virtually zero dead volume; Easy to use; Available in convenient kits tailored specifically to your IC system
Catalog No. NC1761297 Supplier Thermo Scientific Supplier No. 302895
Achieve consistent low dead volume connections; Ensure optimum chromatographic performance for ion chromatography (IC) applications; Conventional PEEK fittings can increase dead volume if the ferrule is incorrectly positioned or tubing poorly cut, resulting in reduced efficiencies and poor peak shapes; Provide worry-free finger-tight connections with virtually zero dead volume; Easy to use; Available in convenient kits tailored specifically to your IC system