Learn More
Thermo Scientific Viper Fingertight Fitting Systems | Stainless Steel | 1 EA
Supplier:  Thermo Scientific 6040.2320
Encompass
- Ferrule-free tightening design for virtually zero-dead-volume connections- Operates at pressures up to 1310 bar- Suitable for temperatures up to 120C- Easy to Install- Fingertight design- Flexible stainless-steel 1/32 in OD- No tools required- Flexible- Compatible with valves and columns from virtually any manufacturer- Fits narrow connections such as 10-port valves
Catalog No. NC2157042