Learn More
Enzo Life Sciences Viperin polyclonal antibody (AT131) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210956C100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa near the C-terminus of human viperin. Source: Purified from rabbit serum. Species reactivity: Human, Mouse. Applications: IP, WB. Recommended Dilutions: Immunoprecipitation (1:200) Western blot (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8195