Learn More
Medchemexpress LLC HY-43869 50mg Medchemexpress, Vipivotide tetraxetan Linker CAS:1703768-74-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY4386950MG
Encompass_Preferred
Medchemexpress, HY-43869 50mg Vipivotide tetraxetan Linker CAS:1703768-74-4 Vipivotide tetraxetan Linker is a useful peptide linker for synthesis of Vipivotide tetraxetan (PSMA-617). Vipivotide tetraxetan (PSMA-617) is a ligand used to synthesize 177 Lu-PSMA-617, which is a radioactive molecule for the treatment of prostate cancer [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2865