Learn More
Eurofins Discoverx PathHunter VIPR2 Total Internalization U2OS Cell Line, 2 vials. (93-0824C3)
Catalog No. 502001369
Encompass
The PathHunter VIPR2 Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of VIPR2 via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged VIPR2 and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the VIPR2 receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1369 Supplier Eurofins Discoverx Supplier No. 930824C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter VIPR2 Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of VIPR2 via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged VIPR2 and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the VIPR2 receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.