Learn More
Cell Biolabs Inc ViraBind™ Adenovirus Miniprep Kit, 10 preps
Catalog No. NC0555700
Encompass_Preferred
Cell Biolabs' ViraBind™ Adenovirus Miniprep Kit provides fast purification of adenovirus supernatant with high yields. The kit uses a special spin column to purify supernatant from a single T75 flask or 10cm dish per prep in about 30 minutes. Total adenovirus recovery is >95%. The kit is suitable for 10 preps.
Catalog No. NC0555700 Supplier Cell Biolabs Inc Supplier No. VPK099
Cell Biolabs' ViraBind™ Adenovirus Miniprep Kit provides fast purification of adenovirus supernatant with high yields. The kit uses a special spin column to purify supernatant from a single T75 flask or 10cm dish per prep in about 30 minutes. Total adenovirus recovery is >95%. The kit is suitable for 10 preps.