Learn More
Alstem ViralBoost Reagent 1mL Research Use Only
SDP
Catalog No. NC0966705
Encompass
Regulates packaging in transcriptional level. Works for both lenti-and retro-viral packaging. Increases viral particle production up to 10-fold. Achieve high titer at low cost. Novel cocktail of small molecules. Easy to use.
Catalog No. NC0966705 Supplier Alstem Supplier No. VB100
Regulates packaging in transcriptional level. Works for both lenti-and retro-viral packaging. Increases viral particle production up to 10-fold. Achieve high titer at low cost. Novel cocktail of small molecules. Easy to use.