Learn More
Cell Biolabs Inc ViraDuctin™ Lentivirus Transduction Kit, 200 transductions
Catalog No. NC9698510
Encompass_Preferred
The rate at which lentiviral vectors bind to and infect cells is mostly controlled by diffusion. During lentivirus infection, only a small fraction of the lentiviral vectors can transduce target cells. Virion adsorption is the limiting step of this process. The use of polycations, such as Polybrene, is standard in many lentiviral infection protocols owing to the observations of improved infection efficiency. Its mechanism of action is thought to involve neutralization of electrostatic repulsion between virion and cell membranes, enhancing attachment. ViraDuctin™ Lentivirus Transduction Kit is a proprietary formulation for the transduction of lentivirus. After forming a complex with lentivirus in culture medium, the ViraDuctin™ reagents typically provide higher transduction efficiency in many cell types compared to reagents such as Polybrene. It is ideal for transduction of nonpermissive cells such as primary cells and stem cells. 200 transductions.
Catalog No. NC9698510 Supplier Cell Biolabs Inc Supplier No. LTV201
The rate at which lentiviral vectors bind to and infect cells is mostly controlled by diffusion. During lentivirus infection, only a small fraction of the lentiviral vectors can transduce target cells. Virion adsorption is the limiting step of this process. The use of polycations, such as Polybrene, is standard in many lentiviral infection protocols owing to the observations of improved infection efficiency. Its mechanism of action is thought to involve neutralization of electrostatic repulsion between virion and cell membranes, enhancing attachment. ViraDuctin™ Lentivirus Transduction Kit is a proprietary formulation for the transduction of lentivirus. After forming a complex with lentivirus in culture medium, the ViraDuctin™ reagents typically provide higher transduction efficiency in many cell types compared to reagents such as Polybrene. It is ideal for transduction of nonpermissive cells such as primary cells and stem cells. 200 transductions.