Learn More
INTACT GENOMICS INC FastAmp Viral and Cell Lysis Solution 100 Reactions (50mL)
Catalog No. 5000082828
Encompass
Intact Genomics' FastAmp Viral and Cell Lysis Solution can be used various sample and cell types stabilizing and prepping DNA or RNA for RPA, subsequent PCR or cDNA synthesis, saving your team significant time and resources.Benefits: No DNA or RNA extraction is needed Safe for sample transport and maintenance Speeds up Covid-19 and other testing processes Compatible with various types of tissues including plants Compatible with different detection technologies Increased efficiency, specificity, and sensitivity Low toxicity to humans/environment Results approved and published by top scientists*
Catalog No. 50-000-82828 Supplier INTACT GENOMICS INC Supplier No. 4631
Intact Genomics' FastAmp Viral and Cell Lysis Solution can be used various sample and cell types stabilizing and prepping DNA or RNA for RPA, subsequent PCR or cDNA synthesis, saving your team significant time and resources.Benefits: No DNA or RNA extraction is needed Safe for sample transport and maintenance Speeds up Covid-19 and other testing processes Compatible with various types of tissues including plants Compatible with different detection technologies Increased efficiency, specificity, and sensitivity Low toxicity to humans/environment Results approved and published by top scientists*