Learn More
Zymo Research Corporation ZR Viral DNA Buffer, 100 ml
SDP
Catalog No. 501251362
Encompass_Preferred
The ZR Viral DNA Buffer is specially formulated buffer for the rapid denaturation and recovery of viral DNA from blood, soft tissues, cultured cells, and biological fluids.
Catalog No. 50-125-1362 Supplier Zymo Research Corporation Supplier No. D30161100
The ZR Viral DNA Buffer is specially formulated buffer for the rapid denaturation and recovery of viral DNA from blood, soft tissues, cultured cells, and biological fluids.