Learn More
Zymo Research Corporation Viral DNA/RNA Buffer, 25 ml
SDP
Catalog No. 502037184
Encompass_Preferred
Viral DNA/RNA Buffer is a reagent that facilitate viral particle lysis and binding of DNA and total RNA from any biological liquid (plasma, serum, urine, blood, saliva, etc.), cellular suspensions and is compatible with samples in DNA/RNA Shield.
Catalog No. 50-203-7184 Supplier Zymo Research Corporation Supplier No. D7020125
Viral DNA/RNA Buffer is a reagent that facilitate viral particle lysis and binding of DNA and total RNA from any biological liquid (plasma, serum, urine, blood, saliva, etc.), cellular suspensions and is compatible with samples in DNA/RNA Shield.