Learn More
Zymo Research Corporation Viral RNA Buffer, 50 ml
SDP
Catalog No. 502037235
Encompass_Preferred
Viral RNA Buffer is a reagent that facilitate viral particle lysis and binding of total RNA from any biological liquid (plasma, serum, urine, blood, saliva, etc.), cellular suspensions and is compatible with samples in DNA/RNA Shield.
Catalog No. 50-203-7235 Supplier Zymo Research Corporation Supplier No. R1034150
Viral RNA Buffer is a reagent that facilitate viral particle lysis and binding of total RNA from any biological liquid (plasma, serum, urine, blood, saliva, etc.), cellular suspensions and is compatible with samples in DNA/RNA Shield.