Learn More
Epoch Life Science Inc GenCatch™ Viral RNA Miniprep Kit (250 preps) Uniquely formulated buffers, reinforced columns with collection tubes, along with protocols.
SDP
Catalog No. 501538101
Encompass_Preferred
Provides a simple, fast and cost-effective method to purify viral nucleic acids from cell-free samples such as plasma, serum, body fluid, urine, and cell culture supernatant. This system utilizes guanidine thiocyanate and 2-mercaptoethanol for their capabilities to inactivate RNase and denature proteins. Purified viral RNA is suitable for applications such as RT-PCR, Northern and RNase protection assays. This system is ideal for isolation of RNA from pathogenic viruses such as hepatitis C virus (HCV) for RT-PCR screening of clinical samples.
Catalog No. 50-153-8101 Supplier Epoch Life Science Inc Supplier No. 1860250
Provides a simple, fast and cost-effective method to purify viral nucleic acids from cell-free samples such as plasma, serum, body fluid, urine, and cell culture supernatant. This system utilizes guanidine thiocyanate and 2-mercaptoethanol for their capabilities to inactivate RNase and denature proteins. Purified viral RNA is suitable for applications such as RT-PCR, Northern and RNase protection assays. This system is ideal for isolation of RNA from pathogenic viruses such as hepatitis C virus (HCV) for RT-PCR screening of clinical samples.