Learn More
Alpco Ltd Viral Stool Panel I RT-PCR Kit
SDP
Catalog No. NC2284613
Encompass
Viral Stool Panel I RT-PCR is a multiplex real-time RT-PCR assay for the direct, qualitative detection and differentiation of norovirus, rotavirus, adenovirus 40/41 and astrovirus in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.
This item is not returnable. View return policy
Catalog No. NC2284613 Supplier Alpco Ltd Supplier No. 86VS1HU100
Viral Stool Panel I RT-PCR is a multiplex real-time RT-PCR assay for the direct, qualitative detection and differentiation of norovirus, rotavirus, adenovirus 40/41 and astrovirus in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.