Learn More
Alpco Ltd Viral Stool Panel II RT-PCR Kit
SDP
Catalog No. NC2284584
Encompass
Viral Stool Panel II RT-PCR is a multiplex real-time RT-PCR assay for the direct, qualitative detection and differentiation of rotavirus, adenovirus 40/41, and astrovirus in human stool samples. This kit is sufficient for 100 tests.
This item is not returnable. View return policy
Catalog No. NC2284584 Supplier Alpco Ltd Supplier No. 86VS2HU100
Viral Stool Panel II RT-PCR is a multiplex real-time RT-PCR assay for the direct, qualitative detection and differentiation of rotavirus, adenovirus 40/41, and astrovirus in human stool samples. This kit is sufficient for 100 tests.