Learn More
Innovative Research Inc Viral Transport Medium 500ml
Catalog No. 502036430
Encompass_Preferred
Viral Transport Medium from Innovative Research has been specially formulated to support ongoing viral research and diagnostics. This preparation follows the standard CDC-recommended formulation and contains 2% FBS,100µg /mL Gentamicin, and 0.5 µg /mL Amphotericin in a Hanks Balanced salt solution base.
This product is suitable for a wide range of purposes, including viral collection, transport, maintenance, and culture. This product can be used for long-term freezer storage, molecular assays, rapid antigen testing, and direct fluorescent antibody (DVA) applications as well.
This is a clear liquid with a pale beige tint and is negative for microbial growth after 48 hours at 37C. Please refer to the lot-specific datasheet that ships with your order for exact osmolarity and pH. This product should be stored at 2-8C and will remain stable for one year from date of manufacture. This product is for research use and is not intended for in vivo use.
For bulk purchas
Catalog No. 50-203-6430 Supplier Innovative Research Inc Supplier No. IGVTM500ML
Viral Transport Medium from Innovative Research has been specially formulated to support ongoing viral research and diagnostics. This preparation follows the standard CDC-recommended formulation and contains 2% FBS,100µg /mL Gentamicin, and 0.5 µg /mL Amphotericin in a Hanks Balanced salt solution base.
This product is suitable for a wide range of purposes, including viral collection, transport, maintenance, and culture. This product can be used for long-term freezer storage, molecular assays, rapid antigen testing, and direct fluorescent antibody (DVA) applications as well.
This is a clear liquid with a pale beige tint and is negative for microbial growth after 48 hours at 37C. Please refer to the lot-specific datasheet that ships with your order for exact osmolarity and pH. This product should be stored at 2-8C and will remain stable for one year from date of manufacture. This product is for research use and is not intended for in vivo use.
For bulk purchas