Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ViraPower™ HiPerform™ Lentiviral Gateway™ Expression Kit

Catalog No. K533000
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K533000 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K533000 Supplier Invitrogen™ Supplier No. K533000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The ViraPower™ HiPerform™ Lentiviral Gateway™ Expression Kit includes all the components needed to generate lentivirus, including vector kit, 293FT cell line, and the support kit.

The ViraPower™ HiPerform™ Lentiviral Gateway™ Expression Kit includes all the components needed to generate lentivirus, including vector kit, 293FT cell line, and the support kit. This kit combines Invitrogen's™ ViraPower™ HiPerform™ Lentiviral and Gateway™ technologies to facilitate easy recombination-based cloning and lentiviral-based, high-level expression of a target gene in dividing and non-dividing mammalian cells. The pLenti6.3/V5-DEST™ vector is equipped with two key genetic elements, making it a HiPerform™ vector: the Woodchuck Posttranscriptional Regulatory Element (WPRE) and the central Polypurine Tract (cPPT) sequence from the HIV-1 integrase gene to produce at least 4-fold increase in protein expression compared to vectors lacking these elements.

Advantages

  • Stable expression
  • Long-term experiments
  • Accurate titer of functional virus
  • Flexible and versatile Gateway™ recombination cloning technology

Key Features

  • HiPerform™ WPRE and cPPT elements
  • CMV promoter
  • V5 epitope tag at C terminus
  • Blasticidin selection

Kit includes

  • pLenti6.3/V5-DEST™ Gateway™ Vector Kit (V533-06)
  • ViraPower™ Bsd Lentiviral Support Kit (K4970-00)
  • 293FT Cell Line (R700-07)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

Vector kit
• pLenti6.3/V5-DEST™ (150 ng/μL), 40 μL, store at -20°C
• pLenti6.3/V5-GW/lacZ Control vector (0.5 μg/μL), 20 μL, store at -20°C
• One Shot™ Stbl3™ Chemically Competent E. coli, 21 x 50 μL, store at -80°C
• pUC19 control DNA (10 pg/μL), 50 μL, store at -80°C
• S.O.C. Medium, 6 mL, store at -80°C

Support kit
• ViraPower™ Packaging Mix, 195 μg, store at -20°C
• Lipofectamine™ 2000, 0.75 mL, store at 4°C
• Blasticidin, 50 mg, store at -20°C
• 293 FT cells (3 x 106 cells/mL), 1 mL, store in liquid nitrogen

Cloning Method Gateway
Constitutive or Inducible System Constitutive
Delivery Type Lentiviral
Cell Type 293-FT
Format Kit
Promoter CMV
Vector pLenti, TOPO-TA Vector
For Use With (Equipment) HiPerform™, TOPO ™ , ViraPower™
For Use With (Application) Protein Expression
Key Functions Viral Expression
Product Line Gateway, HiPerform, ViraPower
Product Type Expression Kit
Protein Tag V5 Epitope Tag
Quantity 1 kit
Selection Agent (Eukaryotic) Blasticidin
Special Elements WPRE, cPPT
Show More Show Less
Can I perform the single-step protocol for the BP/LR Clonase reaction using BP Clonase enzyme and LR Clonase enzyme instead of BP Clonase II enzyme and LR Clonase II enzyme?

In the single-step protocol for the BP/LR Clonase reaction, we would not recommend substituting the BP Clonase II/LR Clonase II enzymes with BP Clonase /LR Clonase enzymes as this would result in very low recombination efficiency.

Do you have a recommended single-step protocol for BP/LR recombination?

Yes, we have come up with a single-step protocol for BP/LR Clonase reaction (http://www.thermofisher.com/us/en/home/life-science/cloning/gateway-cloning.html#1), where DNA fragments can be cloned into Destination vectors in a single step reaction, allowing you to save time and money.

How can I move my gene of interest from a Gateway-adapted expression clone to a new Destination vector as I have lost the entry clone?

We would recommend performing a BP reaction with a Donor vector in order to obtain an entry clone. This entry clone can then be used in an LR reaction with the Destination vector to obtain the new expression clone.

Can I purchase the 5X LR Clonase buffer or 5X BP Clonase buffer separately?

We do not offer the 5X LR Clonase buffer and 5X BP Clonase buffer as standalone products. They are available as part of the enzyme kits.

Do you offer Gateway vectors for expression in plants?

We do not offer any Gateway vectors for expression in plants.

I used one of your lentiviral vectors but am observing cytotoxic effects after transduction. Can you please help?

Possible causes include:

- large volume of viral supernatant used for transduction
- cells sensitive to Polybrene regaent
- too much antibiotic used for selection
- antibiotic used too soon after tranduction
- gene of interest is toxic to cells

I transduced my lentiviral stock into my mammalian cell line but am getting poor expression of my gene of interest. What could have happened?

Poor expression could result from low transduction efficiency, too low of a MOI, too much antibiotic used for selection, usage of antibiotic too soon after transduction, harveting cells too soon after transduction, having a gene of interest that is toxic to cells, or rerrangement in the LTR regions of the expression construct plasmid DNA.

I transduced my lentiviral stock into my mammalian cell line but am getting no expression of my gene of interest. What could have gone wrong?

Here are some possible causes and solutions:
- Promoter silencing; CMV promoter is prone to silencing especially in mouse and rat cells, screen multiple antibiotic resistant clones and select the one with the highest expression levels
- Viral stocks stored incorrectly; aliquot and store at -80 degrees C, do not freeze/thaw more than 3 times

I prepared a lentiviral stock using one of your lentiviral vectors. I am trying to determine the titer using antibiotic selection but am not able to since the cells are very confluent and I am not getting antibiotic-resistant clones. Can you please offer some tips?

Here are some possible causes and solutions:

- Too little antibotic used for selection
- Selection performed on confluent cells; replate cells
- Viral supernatant not diluted sufficiently; titer lentivus using a wider range of 10-fold serial dilutions

I am using one of your lentiviral vectors and am getting a low lentiviral titer. Can you offer some troubleshooting tips?

Possible causes include:

- low transfection efficiency; Use a high-quality plasmid prep, 293FT cells under passage 16, ensure removal of Geneticin during transfection, ensure correct DNA:lipid ratio, and that cells are plated at the correct confluency
- transfected cells are not cultured in medium containing sodium pyruvate; this reagent provides an extra energy source for cells
- viral supernatant harvested too early; viral supernatants can generally be collected 48-72 hrs post-transfection
- viral supernatant too dilute; concentrate virus using CsCl purification
- viral supernatant frozen and thawed multiple times; 3 times should be the maximum freeze/thaw
- gene of interest is large; viral titers decrease as size of insert increases, inserts larger than 5.6 kb are not recommended
- rearrangement in the LTR region of the epxression construct plasmid DNA; use Stb3 cells for transformatin of the lentiviral construct
- poor choice of titering cell line; use HT1080 cells or similar cell line
- Polybrene reagent is not included during transduction; transduce lentiviral construct into cells in the presence of Polybrene reagent
- Lipofectamine reagent handled incorrectly; ensure proper storage and mix gently before use
- Use fluorescence micrscopy to check titer with HiPerform FastTiter lentivirus

My 293FT cells lifted off the plate during lentivirus production. Is this normal?

If 293FT cells detach shortly after transfection (4 hours to overnight):

- This may be a sign of Lipofectamine 2000 toxicity. Cells may have been plated too sparsely prior to transfections.
- The cells may not have been handled gently enough (these cells have a tendency to lift off easily).
- The cells may have been kept at room temperature for too long.

If cells detach 48 to 72 hours post-transfection:
- If the cells lift off in large sheets, this may be a sign of lentivirus production.

I used a mini-prep kit for propagating my lentiviral construct and obtained very low DNA yield. Can you please offer some tips?

The DNA yield from lentiviral mini-prep DNA is often very low due to the presence of the LTRs in the vector backbone. Hence, we do not recommend using a mini-prep kit for propagation of lentiviral constructs. We recommend preparing lentiviral plasmid DNA using the S.N.A.P. MidiPrep Kit (Cat. No. K191001) or the PureLink HiPure Plasmid Midiprep Kit (Cat. No. K210004), both of which contain 10 mM EDTA in the Resuspension Buffer. Since lentiviral DNA midi-preps also often have low DNA yields, we recommend following specific protocols to increase yield- basically, grow cells slowly, use fewer cells per column, and use 100 mL lentivirus culture for each DNA midi-prep.

Note: If you are going to be mini-prepping the lentiviral plasmid during the cloning/colony screening processes, we recommend using the PureLink HQ Mini Kit (Cat. No. K210001) and following the manual protocol with one change: only a single elution with 50 mL TE, pH 8.0 buffer. The typical yield with this method is normally pretty low, 100-150 ng/mL (i.e., 5-7 mg total). The OD 260/280 is typically between 1.8 and 2.1.

I used TOP10 cells for cloning my lentiviral construct. Is that an issue?

We strongly recommend using Stbl3 E. coli for cloning lentiviral constructs. Stbl3 E. coli cells contain the recA13 mutation in their genotype that helps to minimize the likelihood of unwanted recombination between the LTRs. After transforming into Stbl3 E. coli, we recommend picking colonies and validating the lentivirus DNA from mini-preps using Afl II and Xho I digests before proceeding to midi-preps. In all of our lentiviral vectors, Afl II sites are present in both 5' and 3' LTRs, and a Xho I site is present after the 3' end of the MCS. Assuming Afl II cuts only in the LTR sites, and there are no Afl II or Xho I sites in the insert, 3 DNA fragments are expected to be generated from the Afl II + Xho I digest. Any unexpected DNA fragments can be assumed to be a result of LTR recombination. Only clones with the expected pattern of DNA fragments should be chosen for the subsequent midi-prep.

What are the safety features built into the ViraPower Lentiviral Expression System?

The ViraPower Lentiviral Expression System includes the following features designed to enhance its biosafety:

The pLenti expression vector contains a deletion in the 3' LTR (deltaU3) that does not affect the generation of the viral genome in the producer cell line, but results in “self-inactivation” of the lentivirus after transduction of the target cell (References: Yee JK, Moores JC, Jolly DJ, Wolff JA, Respess JG, Friedmann T (1987) Gene Expression from Transcriptionally Disabled Retroviral Vectors. Proc. Natl. Acad. Sci. USA 84: 5197-5201; Yu SF, Ruden Tv, Kantoff PW, Garber C, Seiberg M, Ruther U, Anderson WF, Wagner EF, Gilboa E (1986) Self-Inactivating Retroviral Vectors Designed for Transfer of Whole Genes into Mammalian Cells. Proc. Natl. Acad. Sci. USA 83: 3194-3198; Zufferey R, Dull T, Mandel RJ, Bukovsky A, Quiroz D, Naldini L, Trono D (1998) Selfinactivating Lentivirus Vector for Safe and Efficient in vivo Gene Delivery. J. Virol. 72: 9873-9880). Once integrated into the transduced target cell, the lentiviral genome is no longer capable of producing packageable viral genome.
- The number of genes from HIV-1 used in the system has been reduced to three (i.e., gag, pol, and rev).
- The VSV-G gene from Vesicular Stomatitis Virus is used in place of the HIV-1 envelope (References: Burns JC, Friedmann T, Driever W, Burrascano M, Yee JK (1993) Vesicular Stomatitis Virus G Glycoprotein Pseudotyped Retroviral Vectors: Concentration to a Very High Titer and Efficient Gene Transfer into Mammalian and Nonmammalian Cells. Proc. Natl. Acad. Sci. USA 90: 8033-8037; Emi N, Friedmann T, Yee JK (1991) Pseudotype Formation of Murine Leukemia Virus with the G Protein of Vesicular Stomatitis Virus. J. Virol. 65: 1202-1207; Yee JK, Miyanohara A, LaPorte P, Bouic K, Burns JC, Friedmann T (1994) A General Method for the Generation of High-Titer, Pantropic Retroviral Vectors: Highly Efficient Infection of Primary Hepatocytes. Proc. Natl. Acad. Sci. USA 91: 9564-9568).
- Genes encoding the structural and other components required for packaging the viral genome are separated onto four plasmids. All four plasmids have been engineered not to contain any regions of homology with each other to prevent undesirable recombination events that could lead to the generation of a replication-competent virus (Reference: Dull T, Zufferey R, Kelly M, Mandel RJ, Nguyen M, Trono D, Naldini L (1998) A Third-Generation Lentivirus Vector with a Conditional Packaging System. J. Virol. 72: 8463-8471).
- Although the three packaging plasmids allow in trans expression of proteins required to produce viral progeny (e.g., gal, pol, rev, env) in the 293FT producer cell line, none of them contain LTRs or the psi packaging sequence. This means that none of the HIV-1 structural genes are actually present in the packaged viral genome, and thus, are never expressed in the transduced target cell. No new replication-competent virus can be produced.
- The lentiviral particles produced in this system are replication-incompetent and only carry the gene of interest. No other viral species are produced.' - Expression of the gag and pol genes from pLP1 has been rendered Rev-dependent by virtue of the HIV-1 RRE in the gag/pol mRNA transcript. Addition of the RRE prevents gag and pol expression in the absence of Rev (Reference: Dull T, Zufferey R, Kelly M, Mandel RJ, Nguyen M, Trono D, Naldini L (1998) A Third-Generation Lentivirus Vector with a Conditional Packaging System. J. Virol. 72: 8463-8471).
- A constitutive promoter (RSV promoter) has been placed upstream of the 5' LTR in the pLenti expression vector to offset the requirement for Tat in the efficient production of viral RNA (Reference: Dull T, Zufferey R, Kelly M, Mandel RJ, Nguyen M, Trono D, Naldini L (1998) A Third-Generation Lentivirus Vector with a Conditional Packaging System. J. Virol. 72: 8463-8471).

Despite the presence of the above safety features, the lentivirus produced can still pose some biohazardous risk, since it can transduce primary human cells. For this reason, we highly recommend that you treat lentiviral stocks generated using this system as Biosafety Level 2 (BL-2) organisms and strictly follow all published guidelines for BL-2. Furthermore, exercise extra caution when creating lentivirus carrying potential harmful or toxic genes (e.g., activated oncogenes).
For more information about the BL-2 guidelines and lentivirus handling, refer to the document, “Biosafety in Microbiological and Biomedical Laboratories”, 4th Edition, published by the Centers for Disease Control (CDC) (www.cdc.gov/biosafety/publications/index.htm).

Does the lentivirus produced using your system contain any toxic viral genes?

Lentiviruses produced with our system do not carry or express any viral genes, and therefore have no associated toxicity issues. Only the protein expressed from the coding region between the LTR sites is incorporated into the mammalian cell chromosome and expressed. The lentivirus itself cannot replicate because of the built-in safety features.

Can I leave the lentivirus on the cells after transduction, instead of removing it after an overnight incubation, as my cells are very fragile and have a tendency to slosh off when disturbed?

The concern with leaving the lentivirus on the cells longer would be potential toxicity or growth effects. If you absolutely cannot remove the virus from the cells, we suggest to leave it on the cells and empirically monitor the cells.

What are the physical characteristics of the virus particles?

Note: These are based on the characteristics of HIV.
Morphology: Virions have a complex construction and consist of an envelope, a nucleocapsid, a nucleoid, and a matrix protein. Virions are enveloped, spherical to pleomorphic in shape, and have a size of 80-100 nm in diameter. The surface projections are small or inconspicuous spikes that are densely dispersed, evenly covering the surface. Surface projections are 8 nm long. The core is rod-shaped, or is truncated cone-shaped. The nucleoid is concentric.
Physicochemical and Physical Properties: Virions have a buoyant density in sucrose of 1.13-1.18 g cm-3. Virions are sensitive to treatment with heat, detergents, and formaldehyde. The infectivity is not affected by irradiation.
Proteins: Proteins constitute about 60% of the particle weight. The viral genome encodes structural proteins and non-structural proteins. Virions consist of 5 major structural and 3 non-structural proteins. The virus codes for an RNA-dependent DNA polymerase.
Lipids: Lipids are present and located in the envelope. Virions are composed of 35% lipids by weight. The composition of viral lipids and host cell membranes are similar. The lipids are of host origin, derived from plasma membranes.
Carbohydrates: Three percent of the particle weight is attributed to carbohydrates.

How does Polybrene improve transduction?

Polybrene (hexadimethrine bromide) is a cationic polymer(Cat. No. H9268 from Sigma Aldrich) that increases transduction efficiency by neutralizing the charge repulsion between virus particles and the cell surface. For best results, we recommend performing transduction in the presence of Polybrene.
Note: Some cells (e.g., primary neurons) are sensitive to Polybrene. Hence, before performing a transduction experiment, we recommend testing your cell line for sensitivity to Polybrene at a range of 0-10 µg/mL and avoiding it if the cells exhibit toxicity or phenotypic changes.

What kind of transduction efficiency should I expect to achieve with your ViraPower Lentiviral Expression Systems?

We have found that, in general, 80-90% of the cells in an actively dividing cell line (e.g., ht1080) express a target gene when transduced with lentivirus at an MOI of approximately 1. Some non-dividing cell types transduce lentiviral constructs less efficiently. For example, only about 50% of the cells in a culture of primary human fibroblasts express a target gene when transduced at an MOI of approximately 1. If you are transducing your lentiviral construct into a non-dividing cell type, you may need to increase the MOI (e.g., MOI = 10) to achieve optimal expression levels for your recombinant protein. If you are transducing your lentiviral construct into your mammalian cell for the first time, we recommend using a range of MOIs (e.g., 0, 0.5, 1, 2, 5, 10) to determine the MOI required to obtain the optimal protein expression for your application.

Can I transduce my specific cell type with a lentivirus? How does lentivirus infect cells?

The lentiviral envelope is pseudotyped with the vesicular stomatitis virus glycoprotein (VSV-G), which allows the lentivirus to interact with its target cell in a receptor-independent manner. As a result, lentivirus has broad tropism and can, in theory, transduce any mammalian cell type. This receptor-independent entry into the target cell likely involves endocytosis (Espenshade et al. (2002) Proc Natl Acad Sci U S A 99:11694; Aiken (1997) J Virol 71:5871).

Can I check the titer of my lentivirus by performing PCR/qPCR analysis of viral genes?

You can perform PCR/qPCR analysis of viral genes for lentivirus titering, but keep in mind that the titer will not be a measure of functional virus since it will measure both inactive as well as active virus. As a result, titers obtained using this method are usually about 10-fold higher than with methods that measure functional virus, such as blasticidin selection.

Can I check the titer of my lentivirus using the p24 ELISA assay?

You can use the p24 ELISA assay for lentivirus titering, but keep in mind that the titer will not be a measure of functional virus since it will measure both inactive as well as active virus. As a result, titers obtained using this method are usually about 10-fold higher than with methods that measure functional virus, such as blasticidin selection.

Which cells should I use to titer my lentivirus? Can I use 293, 293FT, or 293T cells?

We strongly recommend the human fibrosarcoma cell line ht1080 Cells (ATCC# CCL-121) as the “gold standard” for titering lentivirus. The primary reason is that transduction efficiency is high in these cells, and titering results are very accurate and reproducible. However, you may also use the same mammalian cell line to titer your lentiviral stocks as you will use to perform your expression studies. In general, this should be an adherent, non-migratory cell line, and exhibit a doubling time in the range of 18-25 hours. Regular 293 cells may be used for lentivirus titering, but we do not recommend using 293T or 293FT cells because these cells contain the SV40 large T antigen that will induce unwanted DNA replication at the SV40 ori contained within the integrated lentiviral expression vector. This often leads to cell death and results in very low titers.Lentivirus is a genus of slow retroviruses, characterized by a long incubation period.

Do you offer a custom service for lentivirus production?

Yes, we do offer a lentivirus production custom service. Please send an email to techsupport@thermofisher.com for details.

After producing lentivirus, can I amplify the virus or do I need to perform another transfection?

Lentivirus produced using our system is replication-incompetent, and this is a safety feature. You must perform a fresh transfection each time you need more virus.

How should I store my lentiviral stocks?

We recommend aliquoting lentiviral stocks immediately after production into small working volumes, and storing at -80 degrees C for long-term storage. Lentivirus is sensitive to storage temperature and to freeze/thaw and should be handled with care. It can lose up to 5% or more activity with each freeze/thaw. When stored properly, viral stocks of an appropriate titer should be suitable for use for up to one year. After long-term storage, we recommend re-titering your viral stocks before use.

Do 293FT cells lift off the plate during lentivirus production?

If 293FT cells detach shortly after transfection (4 hours to overnight):

- This may be a sign of Lipofectamine 2000 toxicity. Cells may have been plated too sparsely prior to transfection.
- The cells may not have been handled gently enough (these cells have a tendency to lift off easily).
- The cells may have been kept at room temperature for too long.

If cells detach 48 to 72 hours post-transfection:
- If the cells lift off in large sheets, this may be a sign of lentivirus production.

What are syncytia?

Syncytia are large multi-nucleated cells that result from VSV-G-induced fusion with neighboring 293FT producer cells. Syncytia production is indicative of high transfection efficiency and lentivirus production. Keep in mind, though, that the absence of syncytia does not mean that virus will not be produced.

Why do I have to culture 293FT cells under Geneticin antibiotic selection?

The 293FT cell line stably expresses the SV40 large T antigen from the pCMVSPORT6Tag.neo plasmid that contains the neomycin resistance marker. In order to maintain the plasmid/phenotype, the cells have to be routinely cultured in medium containing Geneticin (G418) antibiotic at a concentration of 500 µg/mL.

Do you recommend a specific FBS for culturing 293FT cells? Which plastic plates do you recommend?

We use Mycoplasma-tested Gibco FBS (Cat. No. 16000-044). We have observed that when 293FT cells are cultured in the presence of this FBS following the instructions in the manual, virus production is better than that obtained with many other serum sources. We use the following plasticware for 293FT cells:

T175-Fisher Cat. No. 10-126-13; this is a Falcon flask with 0.2 µm vented plug seal cap.
T75-Fisher Cat. No. 07-200-68; this is a Costar flask with 0.2 µm vented seal cap.
100 mm plate-Fisher Cat. No. 08-772E; this is a Falcon tissue culture-treated polystyrene plate.
We get excellent adherence on these plates under routine cell culture/maintenance conditions.

What does the FT stand for in 293FT and why do you recommend using it to produce lentivirus?

The “F” stands for the high transfection efficiency of this particular 293 cell clone (called 293F) and the “T” stands for the SV40 large T antigen. The large T antigen expression plasmid is stably integrated in the genome and confers resistance to Geneticin antibiotic in these cells. The presence of the SV40 large T antigen is important for high-titer lentivirus production and the mechanism is not known. If regular 293 cells or another 293T cell line is used as the producer cell line, you will be able to produce virus, but the titers will be lower.

What is the packaging limit for the lentiviral system?

The size of the wild-type HIV-1 genome is approximately 10 kb. Since the size of the elements required for expression from pLenti vectors adds up to approximately 4-4.4 kb, the size of your gene of interest should theoretically not exceed 5.6-6 kb for efficient packaging. Titers will generally decrease as the size of the insert increases.

Can I purchase the individual plasmids in the ViraPower Lentiviral Packaging Mix?

The ViraPower Lentiviral Packaging Mix consists of an optimized mixture of three viral packaging plasmids, pLP1, pLP2, and pLP VSVG, supplied at 1 mg/mL in TE buffer, pH 8.0. The individual plasmids are not available as standalones.

I have a third-generation lentiviral vector from a different vendor. Will it be compatible with your packaging mix?

Our lentiviral packaging mix belongs to the third generation, meaning that it does not express the tat gene. It can be used with lentiviral vectors that belong to the third generation or higher, where virus production is independent of the tat gene.

I have a second-generation lentiviral vector. Can I use it with your packaging mix?

Your second-generation lentiviral vector will not be compatible with our packaging mix because our packaging mix belongs to the third generation, meaning that it does not express the tat gene; whereas your lentiviral vector will need the tat gene for virus production.

Can I use my own lentiviral packaging mix with one of your lentiviral expression vectors?

Our lentiviral expression vectors belong to the third generation, meaning that they contain a chimeric 5' LTR, by means of which virus production is not dependent on the HIV tat transactivator. As a result, they are compatible with a second- or third-generation packaging mix.

Which generation does your lentiviral packaging mix belong to?

Our lentiviral packaging mix belongs to the third generation, meaning that it does not express the tat gene. Further, gag/pol and rev genes are supplied as independent plasmids, thus eliminating concerns about recombination events bringing components together as a single vector to produce replication-competent lentivirus.

Which generation do your lentiviral expression vectors belong to?

Our lentiviral expression vectors belong to the third generation, meaning that we use a four-plasmid vector system (1 lentiviral expression vector and 3 packaging plasmids), thus eliminating concerns about recombination events bringing components together as a single vector to produce replication-competent lentivirus. Gag/pol and rev genes are supplied as independent plasmids. Further, these vectors contain a chimeric 5' LTR, by means of which virus production is not dependent on the HIV tat transactivator. Also, the original U3 region of the LTR (long terminal repeat) is deleted to make the virus self-inactivating and thus replication-incompetent.

How much of the original viral genome is contained in your lentiviral expression vectors?

Our lentiviral expression vectors contain approximately 20% of the original viral genome. The rest of the viral genome is deleted from our lentiviral expression vectors for safety reasons.

What are the main differences between the ViraPower Lentiviral Expression System, ViraPower HiPerform Lentiviral Expression System, and ViraPower HiPerform Lentiviral FastTiter Expression System?

The main difference between these systems is in the lentiviral expression vector contained within the kits. The ViraPower Lentiviral Expression Vector backbone is similar to the ViraPower HiPerform Lentiviral Expression Vector backbone except that the latter contains two new elements: WPRE (Woodchuck Posttranscriptional Regulatory Element) from the woodchuck hepatitis virus that is placed directly downstream of the gene of interest, allowing for increased transgene expression and the cPPT (central Polypurine Tract) from the HIV-1 integrase gene, which increases the copy number of lentivirus integrating into the host genome and thus allowing for a two-fold increase in viral titer. Together, WPRE and cPPT produce at least a four-fold increase in protein expression in most cell types, compared to the vectors in the ViraPower Lentiviral Expression Systems that do not contain these elements. In the ViraPower HiPerform Fast Titer Expression System, in addition to the WPRE and cPPT elements, the lentiviral expression vector contains the EmGFP reporter gene instead of Bsd, which allows titer of active virus by flow cytometry in just two days post-transduction.

I would like to clone my insert into a lentiviral expression vector. Can my insert have a poly(A) tail? Why are oligo dT-primed cDNA libraries such as SuperScript II premade libraries and their clones not recommended for cloning into pLenti expression vectors?

Inserts cloned into lentiviral vectors should not have a poly(A) signal. The native poly(A) signal (AATAAA or something similar) will be amplified when using the oligo dT during cDNA synthesis. Thus, it will then become part of the cDNA library or its clones.
Since lentivirus is an RNA virus, during the synthesis of the RNA genome to be packaged, if there is a polyadenylation (poly(A)) signal in the insert, the RNA will be terminated prematurely. There is a SV40 poly(A) signal in the vector, but it is after the second LTR, and it is supposed to be there. Almost any clone transferred from a Gateway cDNA library will probably have a poly(A) signal, which, if inserted into a lentiviral vector, would end up terminating the viral RNA prematurely.

In order to circumvent premature termination of the lentiviral RNA, consider these recommendations:
- The desired gene should first be isolated from the library, cloned into an entry vector such as pENTR/D-TOPO without the poly(A) signal (i.e., ATG to Stop), and then transferred into the lentiviral vector.
- If you are trying to establish a lentiviral expression library, you will probably have to go with a library that was amplified using random hexamers rather than an oligo dT, since such a library would be less likely to include a poly(A) signal in the insert.
Size is not usually a problem. The insert size limit of the lentivirus is approximately 5-6 kb (average insert size of the SuperScript II premade libraries is approximately 1.5 kb).

What is the difference between ViraPower lentiviral expression vectors, pLenti6, and pLenti6.2?

In pLenti6 vectors, the blasticidin (Bsd) resistance marker is driven by the SV40 promoter whereas in pLenti6.2 vectors, the Bsd resistance marker is driven by the phosphoglycerate kinase-1 (PGK) promoter.

This difference is important when working with stem cells; the PGK promoter is a native mammalian promoter that is resistant to silencing and shows long-term, persistent expression in stem cells, whereas the SV40 promoter is often silenced over time in primary cells and stem cells.

How do I find out which lentiviral expression vector is in which kit? Why are there so many catalog numbers?

The complete kit composition is listed in the product manual under “Kit Contents and Storage.” Search our website (www.thermofisher.com) using the catalog number to find the product you're interested in. Once you are on the product page, the manual can be viewed and downloaded using the Manuals link. We provide a variety of vectors/kits to suit the various cloning and expression strategies used by different researchers.

Do you offer a control lentiviral expression vector that I can use for checking my transfection and transduction efficiency?

We carry the Vivid Colors pLenti6.3/V5-GW/EmGFP Expression Control Vector (Cat. No. V37006) and Vivid Colors pLenti6.2-GW/EmGFP Expression Control Vector (Cat. No. V36920), both of which are lentiviral vectors containing Emerald Green Fluorescent Protein (EmGFP). They are designed for use with the ViraPower Lentiviral Expression Systems as positive controls to enable the detection of EmGFP fluorescence following transfection in 293FT cells. These vectors serve as titer controls to produce an EmGFP-expressing lentivirus stock and as a transduction control following transduction in both dividing and non-dividing mammalian cells. Both of these vectors are not cloning vectors. The Vivid Colors pLenti6.3/V5-GW/EmGFP Expression Control Vector has the CMV promoter for driving constitutive expression of EmGFP and the PGK promoter for driving long-term, persistent expression of the blasticidin-stable selection marker, whereas the Vivid Colors pLenti6.2-GW/EmGFP Expression Control Vector has the CMV promoter for driving constitutive expression of EmGFP and the SV40 promoter for driving expression of the blasticidin-stable selection marker. In addition, the Vivid Colors pLenti6.3/V5-GW/EmGFP Expression Control Vector is a HiPerform vector, meaning that it is equipped with two key genetic elements: the Woodchuck Posttranscriptional Regulatory Element (WPRE) and the central Polypurine Tract (cPPT) sequence from the HIV-1 integrase gene to produce at least 4-fold increase in increase in protein expression in most cell types, compared to lentiviral vectors that do not contain these elements.

Why do pLenti expression vectors contain an SV40 poly(A) downstream of the 3' LTR, when there is already a poly(A) present in the 3' LTR? Why isn't this poly(A) enough?

All our pLenti expression vectors have 2 poly(A) sites-a poly(A) located within the 3' LTR that is derived from HIV-1 and the SV40 poly(A) downstream of the 3'LTR. The reason for having both is to reduce the chances of transcriptional interference (for instance, if there were a significant amount of transcriptional read-through that continued through the RSV promoter region, this could potentially interfere with transcription from the RSV promoter, which is critical for production of the viral RNA). Once the lentivirus has integrated in the target cells, the SV40 poly(A) will not be present (since the virus just extends from the 5' to 3' LTR), but the poly(A) within the 3' LTR region will still be present and functional.

If I engineer my pLenti expression vector to contain an expression cassette in the reverse orientation with respect to the LTRs, will transcripts arising from the promoter in the reverse orientation interfere with the viral RNA? Will this result in generation of dsRNA that could reduce titers?

The HIV-1 genome consists of two identical copies of single-stranded RNA. Generating dsRNA, as could happen in this instance, will reduce titers since the dsRNA will interfere with genome packaging. Hence, reversing the orientation of the expression cassette with respect to the LTRs will decrease virus titers. Reference: Mautino et al. (2000) Human Gene Therapy 11:895.

Can I use the lentiviral expression vector by itself as an expression vector (without the packaging mix)?

Yes, the lentiviral expression vector will work as an expression vector by itself and can be stably selected with the appropriate antibiotic. Please note that the vector will be about twice the size of most regular vectors. Therefore, you may need to increase the amount of transfected vector to approximate molar equivalents.

Can I use a mini-prep kit for propagation of my lentiviral construct?

We do not recommend using a mini-prep kit for propagation of lentiviral constructs because the DNA yield from lentiviral mini-prep DNA is often very low due to the presence of the LTRs in the vector backbone. We recommend preparing lentiviral plasmid DNA using the S.N.A.P. MidiPrep Kit (Cat. No. K191001) or PureLink HiPure Plasmid Midiprep Kit (Cat. No. K210004), both of which contain 10 mM EDTA in the Resuspension Buffer. Since lentiviral DNA midi-preps also often have low DNA yields, we recommend following specific protocols to increase yield-basically, grow cells slowly, use fewer cells per column, and use 100 mL lentivirus culture for each DNA midi-prep.

Note: If you are going to be mini-prepping the lentiviral plasmid during the cloning/colony screening processes, we recommend using the PureLink HQ Mini Kit (Cat. No. K210001) and following the manual protocol with one change: only a single elution with 50 mL TE, pH 8.0 buffer. The typical yield with this method is normally pretty low: 100-150 ng/mL (i.e., 5-7 mg total). The OD 260/280 is typically between 1.8 and 2.1.

How should I store my lentiviral expression vector?

We recommend storing lentiviral expression vectors at -20 degrees C. Due to their relatively large size, we do not recommend storing these vectors at -80 degrees C, as the vector solution will completely freeze and too many freeze thaws from -80 degrees C will affect the cloning efficiency.

How do I choose between the Gateway and TOPO versions of the lentiviral systems?

The TOPO cloning method is an easy-to-use, 5-minute benchtop PCR cloning method, and we have developed many kits based on this technology. You may want to choose this method if you have only one construct to make. If you plan to place your gene of interest into several different expression systems, you may want to consider Gateway cloning technology, also used in many of our expression kits. Another consideration in choosing the cloning method would be the size of your insert. If your insert is >4 kb, we recommend choosing Gateway cloning, as TOPO cloning will not work efficiently for these large inserts.

What is the backbone for your lentiviral vectors?

Our lentiviral vectors are based on the HIV-1 backbone. However, several alterations have been made so they function solely as a gene delivery vehicle without subsequent viral replication or disease. Specific HIV-1 genes have been deleted to enhance safety. The HIV-1 genes are only expressed in the producer cells (293FT) and none of them are packaged into the viral genome, and thus are never expressed in the transduced target cell.

What type of virus is the lentivirus?

Lentivirus is a genus of slow retroviruses, characterized by a long incubation period.

How do I know whether to choose lentivirus or adenovirus for delivering my gene of interest?

If you are interested in stable integration and selection, choose the lentiviral system. We offer both a Directional TOPO (D-TOPO) and Gateway version of the kit to provide flexibility in the cloning of the gene of interest. If you are looking for transient gene expression, choose the adenoviral system. We offer the Gateway cloning method for this product. Adenoviral vectors can be amplified several times in 293A cells, whereas the only method to concentrate lentivirus is by centrifugation. Adenovirus requires that host cells have th CAR receptor for efficient transduction, whereas due to the VSVG membrane coat on lentivirus particles, these viruses have broad tropism for a variety of mammalian cell types.

It should be noted, however, that gene expression from both systems is typically detected within 24-48 hours of transduction, so both systems can be used for experiments of a transient nature. The main difference is that lentivirus integrates into the host genome and adenovirus does not. Higher viral titers are achieved with the adenovirus.

What is MOI, and how do I know which MOI to use?

MOI stands for multiplicity of infection. Theoretically, an MOI of 1 will provide 1 virus particle for each cell on a plate, while an MOI of 10 represents ten virus particles per cell. However, several factors can influence the optimal MOI including the nature of your mammalian cell line, (non-dividing vs. dividing), transduction efficiency, your application of interest, and your protein of interest.

When transducing your adenoviral or lentiviral construct into the mammalian cell line of choice for the first time, we recommend using a range of MOIs (0, 0.5, 1, 2, 5, 10, 50) to determine the MOI required to obtain optimal gene expression. MOIs greater than 50, such as MOI 100, are common for the transduction of neurons with lentivirus. After you determine the MOI that gives optimal gene expression, subsequent transductions can be performed at the optimal MOI.

How do I decide between adenoviral and lentiviral expression systems?

Adenoviral expression is used for transient expression, whereas lentiviral expression is used for longer-term expression. Adenoviral vectors can be amplified several times in 293A cells, whereas the only method to concentrate lentivirus is by centrifugation. Adenovirus requires that host cells have the CAR receptor for efficient transduction, whereas due to the VSVG membrane coat on lentivirus particles, these viruses have broad tropism for a variety of mammalian cell types.

Do I need to include a consensus Kozak sequence when I clone my gene of interest into one of your mammalian expression vectors?

The consensus Kozak sequence is A/G NNATGG, where the ATG indicates the initiation codon. Point mutations in the nucleotides surrounding the ATG have been shown to modulate translation efficiency. Although we make a general recommendation to include a Kozak consensus sequence, the necessity depends on the gene of interest and often, the ATG alone may be sufficient for efficient translation initiation. The best advice is to keep the native start site found in the cDNA unless one knows that it is not functionally ideal. If concerned about expression, it is advisable to test two constructs, one with the native start site and the other with a consensus Kozak. In general, all expression vectors that have an N-terminal fusion will already have an initiation site for translation.

Do I need to include a ribosomal binding site (RBS/Shine Dalgarno sequence) or Kozak sequence when I clone my gene of interest?

ATG is often sufficient for efficient translation initiation although it depends upon the gene of interest. The best advice is to keep the native start site found in the cDNA unless one knows that it is not functionally ideal. If concerned about expression, it is advisable to test two constructs, one with the native start site and the other with a Shine Dalgarno sequence/RBS or consensus Kozak sequence (ACCAUGG), as the case may be. In general, all expression vectors that have an N-terminal fusion will already have a RBS or initiation site for translation.

What is the composition of the ViraPower lentiviral packaging mix?

The ViraPower lentiviral packaging mix is an optimized mix of three packaging plasmids: pLP1, pLP2 and pLP/VSVG, supplied in TE buffer, pH 8.0, at a concentration of 1 µg/µL. These plasmids supply the helper functions as well as structural and replication proteins in trans that are required to produce lentivirus using our ViraPower lentiviral expression vectors.

What is Polybrene and how does it facilitate lentiviral transduction?

Polybrene (hexadimethrine bromide) is a cationic polymer used to increase the efficiency of infection of certain cells with virus particles in cell culture. Polybrene acts by neutralizing the charge repulsion between the virions and cell surface.

How should I culture my 293FT cells?

The plasmid expressing the SV40 large T antigen, pCMVSPORT6TAg.neo, is stably integrated in 293FT cells. Hence, we recommend culturing 293FT cells in the presence of 500 µg/mL Geneticin (G418) antibiotic.

What are the safety features that have been engineered into the Virapower lentiviral expression system?

Here is a summary of the safety features that have been built into the ViraPower lentiviral expression system.
• The system is a 3rd generation system with regard to safety features. Our lentiviral expression vectors are derived from wild-type HIV, but nearly all the wild type viral proteins (e.g., Vpr, Vpu, Vif, Nef, Tat) have been removed and the HIV envelope is not used. VSV-G (vesicular stomatitis virus G) envelope protein is used instead.
• The system is a four-plasmid system, and the packaging functions are supplied in the packaging mix in trans, eliminating concerns about recombination events bringing components together as a single vector.
• The lentivirus particles that are generated carry only the gene of interest, and the packaging mix vectors never become part of the packaged virus.
• The lentivirus has a self-inactivation feature, meaning that once it is transduced into cells and has integrated into the mammalian cell host, it is no longer capable of producing a packageable viral genome.
• Expression of gag/pol from pLP1 is rev dependent, so both plasmids (pLP1 and pLP2) must be co-expressed to package the gene of interest into virions.

What safety precautions should I take for use of the ViraPower lentiviral expression system?

There are many safety features that we have built into our ViraPower lentiviral expression system. We recommend strict adherence to all CDC guidelines for Biosafety Level 2 (BL-2), as well as institutional guidelines. BL-2 guidelines may be found in “Biosafety in Microbiological and Biomedical Laboratories” 4th edition, Centers for Disease Control (www.cdc.gov).

How should I store my lentivirus?

Lentivirus should be aliquoted immediately after production and stored at -80 degrees C. Lentivirus can lose up to 5% of titer with each freeze/thaw. When stored properly, viral stocks should be suitable for use for up to one year. After long-term storage, we recommend re-titering the viral stocks before use.

What is the insert size limit for ViraPower lentiviral vectors?

As the size of the insert increases, lentiviral titers decrease. The size of the wild-type HIV genome is approximately 10 kb. Since the size of the elements required for expression from ViraPower lentiviral vectors totals approximately 3–4.5 kb, the size of the insert should not exceed 5.6–6 kb. In-house data shows that viral titer drops approximately 2-fold for every kb of insert size over 4 kb. If you wish to produce lentivirus with an insert of > 4 kb, you will need to concentrate the virus to obtain a suitable titer.

How can I increase lentiviral transduction efficiency?

a. Try different MOIs. All cells are not created equal.
b. Try Polybrene. Test in the range 0–10 µg/mL (manual recommends 6 µg/mL).
c. If Polybrene does not help, you might try using DEAE-dextran. We have not tried it in-house, but it has been used in the literature [Reiser et al. (1996) PNAS 93, 15271].

How do I determine lentiviral transduction efficiency?

ViraPower lentiviral expression kits contain a control lentiviral expression vector (the lacZ- expression vector or GFP- expression vector). When packaged into virions and transduced into a mammalian cell line, these vectors express either ß-galactosidase or GFP, respectively. If you use the lacZ control virus, you can use the ß-galactosidase staining kit (Cat. No. K1455-01) to determine transduction efficiency. If you use the GFP control virus, you can use a fluorescent microscope or perform FACS analysis.

Can you tell me the difference between a Shine-Dalgarno sequence and a Kozak sequence?

Prokaryotic mRNAs contain a Shine-Dalgarno sequence, also known as a ribosome binding site (RBS), which is composed of the polypurine sequence AGGAGG located just 5’ of the AUG initiation codon. This sequence allows the message to bind efficiently to the ribosome due to its complementarity with the 3’-end of the 16S rRNA. Similarly, eukaryotic (and specifically mammalian) mRNA also contains sequence information important for efficient translation. However, this sequence, termed a Kozak sequence, is not a true ribosome binding site, but rather a translation initiation enhancer. The Kozak consensus sequence is ACCAUGG, where AUG is the initiation codon. A purine (A/G) in position -3 has a dominant effect; with a pyrimidine (C/T) in position -3, translation becomes more sensitive to changes in positions -1, -2, and +4. Expression levels can be reduced up to 95% when the -3 position is changed from a purine to pyrimidine. The +4 position has less influence on expression levels where approximately 50% reduction is seen. See the following references:

- Kozak, M. (1986) Point mutations define a sequence flanking the AUG initiator codon that modulates translation by eukaryotic ribosomes. Cell 44, 283-292.
- Kozak, M. (1987) At least six nucleotides preceding the AUG initiator codon enhance translation in mammalian cells. J. Mol. Biol. 196, 947-950.
- Kozak, M. (1987) An analysis of 5´-noncoding sequences from 699 vertebrate messenger RNAs. Nucleic Acids Res. 15, 8125-8148.
- Kozak, M. (1989) The scanning model for translation: An update. J. Cell Biol. 108, 229-241.
- Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res. 18, 2828.

Note: The optimal Kozak sequence for Drosophila differs slightly, and yeast do not follow this rule at all. See the following references:

- Romanos, M.A., Scorer, C.A., Clare, J.J. (1992) Foreign gene expression in yeast: a review. Yeast 8, 423-488.
- Cavaneer, D.R. (1987) Comparison of the consensus sequence flanking translational start sites in Drosophila and vertebrates. Nucleic Acids Res. 15, 1353-1361.

Do you offer pLenti6.3/V5-DEST Gateway vector as a stand-alone product?

Unfortunately, we do not offer the pLenti6.3/V5-DEST Gateway vector as a stand-alone product. The pLenti6.3/V5-DEST Gateway vector is offered as a component of the pLenti6.3/V5-DEST Gateway Vector Kit (Cat. No. V53306) or as part of the ViraPower HiPerform Lentiviral Gateway Expression Kit (Cat. No. K5330-00).

What is the consensus Kozak sequence and what is the function of the Kozak sequence?

Eukaryotic (and specifically mammalian) mRNA contains sequence information that is important for efficient translation. However, this sequence, termed a Kozak sequence, is not a true ribosome binding site, but rather a translation initiation enhancer. The Kozak consensus sequence is ACCAUGG, where AUG is the initiation codon. A purine (A/G) in position -3 has a dominant effect; with a pyrimidine (C/T) in position -3, translation becomes more sensitive to changes in positions -1, -2, and +4. Expression levels can be reduced up to 95% when the -3 position is changed from a purine to pyrimidine. The +4 position has less influence on expression levels where approximately 50% reduction is seen. See the following references:

Kozak, M. (1986) Point mutations define a sequence flanking the AUG initiator codon that modulates translation by eukaryotic ribosomes. Cell 44, 283-292.
Kozak, M. (1987) At least six nucleotides preceding the AUG initiator codon enhance translation in mammalian cells. J. Mol. Biol. 196, 947-950.
Kozak, M. (1987) An analysis of 5´-noncoding sequences from 699 vertebrate messenger RNAs. Nucleic Acids Res. 15, 8125-8148.
Kozak, M. (1989) The scanning model for translation: An update. J. Cell Biol. 108, 229-241.
Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res. 18, 2828.

Note: The optimal Kozak sequence for Drosophila differs slightly, and yeast do not follow this rule at all. See the following references:

Romanos, M.A., Scorer, C.A., Clare, J.J. (1992) Foreign gene expression in yeast: a review. Yeast 8, 423-488.
Cavaneer, D.R. (1987) Comparison of the consensus sequence flanking translational start sites in Drosophila and vertebrates. Nucleic Acids Res. 15, 1353-1361.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.