Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ViraPower™ HiPerform™ T-REx™ Gateway™ Expression System

Catalog No. A11141
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11141 20 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A11141 Supplier Invitrogen™ Supplier No. A11141
Add to Cart
Add to Cart
Request a Quote

The ViraPower™ HiPerform™ T-REx™ Gateway™ Expression System includes all the components needed to generate lentivirus, including vector kit, 293FT cell line, and the support kit.

The ViraPower™ HiPerform™ T-REx™ Gateway™ Expression System includes all the components needed to generate lentivirus, including vector kit, 293FT cell line, and the support kit. This kit combines Invitrogen's ViraPower™ HiPerform™ Lentiviral, T-REx™ and Gateway™ technologies to facilitate easy recombination-based cloning and lentiviral-based, regulated (Tetracycline-inducible), high-level expression of a target gene in dividing and non-dividing mammalian cells. The pLenti6.3/ TO/V5-DEST vector is equipped with two key genetic elements, making it a HiPerform™ vector: the Woodchuck Posttranscriptional Regulatory Element (WPRE) and the central Polypurine Tract (cPPT) sequence from the HIV-1 integrase gene to produce at least 4-fold increase in protein expression compared to vectors lacking these elements.

Advantages

  • Generates replication-incompetent lentivirus for transducing dividing and non-dividing mammalian cells
  • Easy recombination-based cloning using Gateway™ technology
  • Stable, long-term, tetracycline-regulated expression
  • Enhanced protein expression, up to 4-fold or greater, compared to traditional lentiviral expression systems

Key Features

  • WPRE (Woodchuck Posttranscriptional Regulatory Element) from the woodchuck hepatitis virus, increases transgene expression and cPPT (central Polypurine Tract) from the HIV-1 integrase gene, increases the copy number of lentivirus integrating into the host genome, thus increasing viral titer. WPRE and cPPT together produce at least a four-fold increase in protein expression in most cell types, compared to other vectors that do not contain these elements.
  • Hybrid promoter consisting of the human cytomegalovirus (CMV) promoter and two tetracycline operator 2 (TetO2) sites for high-level, regulated expression of the target gene
  • Blasticidin selection marker for stable selection under control of SV40 promoter

Kit includes

  • ViraPower™ HiPerform™ T-Rex™ Gateway™ Vector Kit (Cat. No. A11144)
  • ViraPower™ Bsd Lentiviral Support Kit (K497000)
  • 293FT Cell Line (R70007)
  • Gateway™ LR Clonase™ II Plus Enzyme Mix (12538120)
  • One Shot™ Stbl3™ Chemically Competent E. coli (C737303)
  • Geneticin™ (10131035)
  • pENTR™ Gus positive control plasmid
TRUSTED_SUSTAINABILITY

Specifications

Antibiotic Resistance Bacterial Ampicillin (AmpR), Blasticidin (BsdR)
Content And Storage

ViraPower™ HiPerform™ T-Rex™ Gateway™ Vector Kit (Cat. No. A11144)
• pLenti6.3/TO/V5-DEST vector (40 μL of vector at 150 ng/μL in TE Buffer, pH 8.0), 6 μg
• pLenti3.3/TR vector (40 μL of vector at 0.5 μg/μL in TE Buffer, pH 8.0), 20 μg
• pLenti6.3/TO/V5-GW/lacZ plasmid (20 μL of plasmid at 0.5 μg/μL in TE Buffer, pH 8.0), 10 μg
• Tetracycline (10 mg/mL in water), 1 mL. Store at -20°C, protect from light.
Store at -20°C unless otherwise noted

ViraPower™ Bsd Lentiviral Support Kit (Cat. No. K497000)
• ViraPower™ Lentiviral Packaging Mix (Contains 195 μL of a mixture of pLP1, pLP2, and pLP/VSVG plasmids, at 1 μg/μL in TE Buffer, pH 8.0), 195 μg. Store at -20°C.
• Lipofectamine™ 2000 (Cat. No. 11668027), 0.75 mL. Store at 4°C (Do not freeze)
• Blasticidin powder (Cat. No. R21001), 50 mg. Store at -20°C.

293FT Cell Line (Cat. No. R70007)
• 3 x 106 frozen cells in 1 mL of Freezing Medium, 1 vial. Store in liquid nitrogen

Gateway™ LR Clonase™ II Plus Enzyme Mix (Cat. No. 12538120)
• Gateway™ LR Clonase™ II Plus Enzyme Mix, 40 μL. Store -20°C for up to 6 months, -80°C for long term storage.
• Proteinase K Solution (2 ug/mL), 40 μL

One Shot™ Stbl3™ Chemically Competent E. coli (Cat. No. C737303)
• Stbl3™ Cells, 6 mL of S.O.C. Medium, 21 x 50 μL. Store at -80°C.

Geneticin™ (Cat. No. 10131035)
• Geneticin™ solution (50 mg/mL), 20 mL, store at 4°C or -20°C
• pENTR™ Gus positive control plasmid, store at -20°C

pUC19 control DNA, (10 pg/μL), 50 μL

Cloning Method Gateway
Constitutive or Inducible System Inducible
Delivery Type Lentiviral
Cell Type 293-FT
Format Kit
Promoter CMV/TO
Vector pLenti
Gateway Enzyme LR Clonase™ II Plus, LR Clonase™ II
For Use With (Equipment) Gateway™, HiPerform™, T-REx™, ViraPower™
For Use With (Application) Protein Expression
Inducing Agent Doxycycline, Tetracycline
Key Functions Viral Expression
No. of Reactions 20 Reactions
PCR Enzyme Proof-Reading Polymerase
Product Line Gateway, HiPerform, T-REx, ViraPower
Product Type Mammalian Expression system
Protein Tag V5 Epitope Tag
Quantity 20 Reactions
Selection Agent (Eukaryotic) Blasticidin
Replication Origin pUC Origin
Selection Marker Eukaryotic BsdR
Selection Marker Promoter PGK Promoter
Special Elements WPRE, cPPT
Show More Show Less
Can I perform the single-step protocol for the BP/LR Clonase reaction using BP Clonase enzyme and LR Clonase enzyme instead of BP Clonase II enzyme and LR Clonase II enzyme?

In the single-step protocol for the BP/LR Clonase reaction, we would not recommend substituting the BP Clonase II/LR Clonase II enzymes with BP Clonase /LR Clonase enzymes as this would result in very low recombination efficiency.

Do you have a recommended single-step protocol for BP/LR recombination?

Yes, we have come up with a single-step protocol for BP/LR Clonase reaction (http://www.thermofisher.com/us/en/home/life-science/cloning/gateway-cloning.html#1), where DNA fragments can be cloned into Destination vectors in a single step reaction, allowing you to save time and money.

How can I move my gene of interest from a Gateway-adapted expression clone to a new Destination vector as I have lost the entry clone?

We would recommend performing a BP reaction with a Donor vector in order to obtain an entry clone. This entry clone can then be used in an LR reaction with the Destination vector to obtain the new expression clone.

Can I purchase the 5X LR Clonase buffer or 5X BP Clonase buffer separately?

We do not offer the 5X LR Clonase buffer and 5X BP Clonase buffer as standalone products. They are available as part of the enzyme kits.

Do you offer Gateway vectors for expression in plants?

We do not offer any Gateway vectors for expression in plants.

I am using your ViraPower HiPerform T-REx Gateway Expression System and am getting high basal level expression of my gene of interest in transient co-transduction experiments. Can you please offer some suggestions?

Here are some possible causes and solutions:

- Transduced Lenti3.3/TR viral construct at too low of an MOI when compared to the epxression construct; Transduce the Lenti3.3TR viral construct into mammalian cells at a higher MOI (e.g., MOI of 10) than the expression construct (e.g., MOI 1-5)
- Did not wait for a sufficient amount of time after tranducing the Lenti3.3/TR viral construct before transducing the Lenti6.3/TO/V5-DEST viral construct; Transduce mammalian cells with the Lenti6/TR construct, and then wait for 24 hrs before tranducing cells with the Lenti6.3/TO/V5-DEST construct.

I am using your ViraPower HiPerform T-REx Gateway Expression System and am getting high basal level expression of my gene of interest. Can you please offer some suggestions?

This can happen if the Lenti6.3/TO/V5-DEST construct is transduced into cells that do not express the Tet repressor. Generate a ViraPower T-REx cell line first, and then use this cell line as the host for the Lenti6.3/TO/V5-DEST virus. You may also want to confirm that your cells are not being grown in medium with FBS that has high tetracycline levels in it.

I am using your ViraPower HiPerform T-REx Gateway Expression System. I get very poor expression of my gene of interest. Can you please offer some tips?

Here are some possible causes and solutions:

- Cells harvested and assayed too soon after addition of tetracycline; culture cells for a longer period of time after addition of tetracycline before assaying for recombinant protein epxression (at least 24 hrs)
- Lenti6.3/TO/V5-DEST lentiviral stock not titered; Titer the stock (see page 21 of the manual)
- Lenti6.3/TO/V5-DEST lentiviral stock stored incorrectly; Aliquote and store stocks at -80 degrees C, do not freeze/thaw more than 3 times; re-titer stock before use if stored longer than 6 months

I am using your ViraPower HiPerform T-REx Gateway Expression System and am seeing very low levels of Tet repressor expressed. What should I do?

Here are some possible causes and solutions:

- Lenti3.3/TR construct integrated into an inactive region of the genome; Screen other Geneticin-resistant colonies and choose the clone that exhibits the highest level of Tet repressor expression for use as the hose for your Lenti6.3/TO/V5-DEST construct.
- Tranduced Lenti3.3/TR into a mammalian cell line in which the CMV promoter is downregulated; use another mammalian cell line for transduction.

Do you carry a lentiviral expression system for inducible/regulated expression of my gene of interest?

We offer the ViraPower HiPerform T-REx Gateway Expression System (Cat. No. A11141) that combines ViraPower HiPerform Lentiviral and T-REx technologies to facilitate lentiviral-based, regulated, high-level expression of a target gene in dividing or non-dividing mammalian cells. The ViraPower HiPerform T-REx Gateway Vector Kit is available separately (Cat. No. A11144).

I performed stable selection but my antibiotic-resistant clones do not express my gene of interest. What could have gone wrong?

Here are possible causes and solutions:

Detection method may not be appropriate or sensitive enough:
- We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot.
- Insufficient number of clones screened: Screen at least 20 clones.
- Inappropriate antibiotic concentration used for stable selection: Make sure the antibiotic kill curve was performed correctly. Since the potency of a given antibiotic depends upon cell type, serum, medium, and culture technique, the dose must be determined each time a stable selection is performed. Even the stable cell lines we offer may be more or less sensitive to the dose we recommend if the medium or serum is significantly different.
- Expression of gene product (even low level) may not be compatible with growth of the cell line: Use an inducible expression system.
- Negative clones may result from preferential linearization at a vector site critical for expression of the gene of interest: Linearize the vector at a site that is not critical for expression, such as within the bacterial resistance marker.

I used a mammalian expression vector but do not get any expression of my protein. Can you help me troubleshoot?

Here are possible causes and solutions:

- Try the control expression that is included in the kit
Possible detection problem:

- Detection of expressed protein may not be possible in a transient transfection, since the transfection efficiency may be too low for detection by methods that assess the entire transfected population. We recommend optimizing the transfection efficiency, doing stable selection, or using methods that permit examination of individual cells. You can also increase the level of expression by changing the promoter or cell type.
- Expression within the cell may be too low for the chosen detection method. We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot. Protein might be degraded or truncated: Check on a Northern. Possible time-course issue: Since the expression of a protein over time will depend upon the nature of the protein, we always recommend doing a time course for expression. A pilot time-course assay will help to determine the optimal window for expression. Possible cloning issues: Verify clones by restriction digestion and/or sequencing.

I am using a mammalian expression vector that has the neomycin resistance gene. Can I use neomycin for stable selection in mammalian cells?

No; neomycin is toxic to mammalian cells. We recommend using Geneticin (a.k.a. G418 Sulfate), as it is a less toxic and very effective alternative for selection in mammalian cells.

Is it okay if my construct has an ATG that is upstream of the ATG in my gene of interest? Will it interfere with translation of my gene?

Translation initiation will occur at the first ATG encountered by the ribosome, although in the absence of a Kozak sequence, initiation will be relatively weak. Any insert downstream would express a fusion protein if it is in frame with this initial ATG, but levels of expressed protein are predicted to be low if there is a non-Kozak consensus sequence. If the vector contains a non-Kozak consensus ATG, we recommend that you clone your gene upstream of that ATG and include a Kozak sequence for optimal expression.

Why is sequential transfection recommended over co-transfection in the T-REx and GeneSwitch systems?

When a co-transfection is performed, there is no way of testing the double stable cell line for functional TetR or GeneSwitch protein, respectively. On the other hand, when sequential transfection is performed, one can functionally test the generated T-REx or GeneSwitch cell line by transiently transfecting the lacZ expression control plasmid and then picking a clone that shows the lowest basal level of expression of lacZ in the absence of the inducer, and the highest level of lacZ in the presence of the inducer. This clone can then be expanded and used to transfect the T-REx or GeneSwitch expression construct, as the case may be.

What is the main advantage of the GeneSwitch system over the T-REx system? And what is its main disadvantage?

With the GeneSwitch system, it is possible to have the absolute lowest basal levels of expression of the gene of interest, whereas the T-REx system may be a little leaky due to the inevitable presence of tetracycline in FBS. The induced level of expression in the GeneSwitch system can be even higher than that seen with the CMV promoter. The disadvantage of the GeneSwitch system is that the expression does not appear to switch off very easily in culture, although it has been demonstrated to function beautifully in transgenics. The T-REx system, on the other hand, can be switched on and off by the addition and removal of the inducer.

What is the advantage of the Flp-In T-REx system over the T-REx system?

The Flp-In T-REx system combines the targeted integration offered by the Flp-In system with the powerful inducible expression offered by the T-REx system. It allows generation of isogenic, inducible, stable cell lines and permits polyclonal selection of these cell lines. Once the Flp-In T-REx host cell line containing an integrated FRT site has been created, subsequent generation of Flp-In T-REx cell lines expressing the gene(s) of interest is rapid and efficient.

Can I use doxycycline instead of tetracycline as an inducer in the T-REx system?

Doxycycline may be used as an alternative inducing agent in the T-REx system. It is similar to tetracycline in its mechanism of action, and exhibits similar dose-response and induction characteristics as tetracycline in the T-REx system. Doxycycline has been shown to have a longer half-life than tetracycline (48 hours vs. 24 hours, respectively). We do not offer doxycycline, but it may be obtained from Sigma (Cat. No. D9891).

I am interested in a mammalian expression system where I can have regulated expression of my gene of interest. I see that you offer multiple systems for this purpose. Can you describe the main features of each system?

We offer three unique mammalian expression systems for inducible/regulated expression of the gene of interest:

- T-REx system
- Flp-In T-REx system
- GeneSwitch system

Please see below to see how they compare with one another:
System -- Basal Expression Level -- Induced Expression Level -- Response time to Maximal Expression -- Transgenic Appliation
T-Rex system -- Low -- Highest -- High -- Suitable
Flp-In T-REx system -- Lower -- High -- 24-48 hrs -- Suitable
GeneSwitch system -- Lowest -- High -- 24-48 hrs -- Suitable

Do you offer a GFP-expressing mammalian expression vector that I can use as a control to monitor my transfection and expression?

We offer pJTI R4 Exp CMV EmGFP pA Vector, Cat. No. A14146, which you can use to monitor your transfection and expression.

Which competent E. coli do you recommend using for propagation of my Gateway-adapted mammalian Destination vector?

We recommend using One Shot ccdB Survival 2 T1R Competent Cells, Cat. No. A10460. This strain is resistant to the toxic effects of the ccdB gene. Note: Do not use general E. coli cloning strains, including TOP10 or DH5alpha, for propagation and maintenance, as these strains are sensitive to ccdB effects.

I am working with a mouse cell line and would like to express my gene at high levels using one of your vectors with the CMV promoter. Do you foresee any problems with this approach?

The CMV promoter is known to be downregulated over time in mouse cell lines. Hence, we recommend using one of our non-CMV vectors, such as those with the EF1alpha or UbC promoter, for long-term expression in mouse cell lines.

Do I need to include a consensus Kozak sequence when I clone my gene of interest into one of your mammalian expression vectors?

The consensus Kozak sequence is A/G NNATGG, where the ATG indicates the initiation codon. Point mutations in the nucleotides surrounding the ATG have been shown to modulate translation efficiency. Although we make a general recommendation to include a Kozak consensus sequence, the necessity depends on the gene of interest and often, the ATG alone may be sufficient for efficient translation initiation. The best advice is to keep the native start site found in the cDNA unless one knows that it is not functionally ideal. If concerned about expression, it is advisable to test two constructs, one with the native start site and the other with a consensus Kozak. In general, all expression vectors that have an N-terminal fusion will already have an initiation site for translation.

Do I need to include a ribosomal binding site (RBS/Shine Dalgarno sequence) or Kozak sequence when I clone my gene of interest?

ATG is often sufficient for efficient translation initiation although it depends upon the gene of interest. The best advice is to keep the native start site found in the cDNA unless one knows that it is not functionally ideal. If concerned about expression, it is advisable to test two constructs, one with the native start site and the other with a Shine Dalgarno sequence/RBS or consensus Kozak sequence (ACCAUGG), as the case may be. In general, all expression vectors that have an N-terminal fusion will already have a RBS or initiation site for translation.

Can the Clontech Tet-On system or Tet-Off system components be used with your T-REx tetracycline-regulated mammalian expression system?

No. The two systems are not compatible since they utilize different strategies for promoter regulation. The T-REx system is designed such that native E. coli tet-repressor protein molecules bind to specific tet-operator sequences (2X TO) just downstream of the TATA box in the full length CMV promoter in the expression vector. This binding keeps the promoter silent simply by preventing the normal transcription machinery from productive assembly at the TATA box. Incidentally, it is this full length CMV promoter region that permits higher induced expression levels relative to other systems.

The recombinant 'repressor' proteins utilized in Clontech's system are actually recombinant fusion proteins which also contain a potent transcriptional transactivator. The Clontech system places operator sequences 5' to the TATA box and relies upon the VP16 transactivator to promote transcription. These repressor-transactivator fusion constructs would have unpredictable and unreliable effects at the CMV promoter in our expression constructs. Additionally, the tet-repressor protein produced from the pCDNA6/TR construct in the T-REx system has no transactivation domain and so would exert little regulatory effect at the minimal promoter region (non-full length CMV) found in the Clontech response plasmids.

Can you tell me the difference between a Shine-Dalgarno sequence and a Kozak sequence?

Prokaryotic mRNAs contain a Shine-Dalgarno sequence, also known as a ribosome binding site (RBS), which is composed of the polypurine sequence AGGAGG located just 5’ of the AUG initiation codon. This sequence allows the message to bind efficiently to the ribosome due to its complementarity with the 3’-end of the 16S rRNA. Similarly, eukaryotic (and specifically mammalian) mRNA also contains sequence information important for efficient translation. However, this sequence, termed a Kozak sequence, is not a true ribosome binding site, but rather a translation initiation enhancer. The Kozak consensus sequence is ACCAUGG, where AUG is the initiation codon. A purine (A/G) in position -3 has a dominant effect; with a pyrimidine (C/T) in position -3, translation becomes more sensitive to changes in positions -1, -2, and +4. Expression levels can be reduced up to 95% when the -3 position is changed from a purine to pyrimidine. The +4 position has less influence on expression levels where approximately 50% reduction is seen. See the following references:

- Kozak, M. (1986) Point mutations define a sequence flanking the AUG initiator codon that modulates translation by eukaryotic ribosomes. Cell 44, 283-292.
- Kozak, M. (1987) At least six nucleotides preceding the AUG initiator codon enhance translation in mammalian cells. J. Mol. Biol. 196, 947-950.
- Kozak, M. (1987) An analysis of 5´-noncoding sequences from 699 vertebrate messenger RNAs. Nucleic Acids Res. 15, 8125-8148.
- Kozak, M. (1989) The scanning model for translation: An update. J. Cell Biol. 108, 229-241.
- Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res. 18, 2828.

Note: The optimal Kozak sequence for Drosophila differs slightly, and yeast do not follow this rule at all. See the following references:

- Romanos, M.A., Scorer, C.A., Clare, J.J. (1992) Foreign gene expression in yeast: a review. Yeast 8, 423-488.
- Cavaneer, D.R. (1987) Comparison of the consensus sequence flanking translational start sites in Drosophila and vertebrates. Nucleic Acids Res. 15, 1353-1361.

I sequenced one of your vectors after PCR amplification and observed a difference from what is provided online (or in the manual). Should I be concerned?

Our vectors have not been completely sequenced. Your sequence data may differ when compared to what is provided. Known mutations that do not affect the function of the vector are annotated in public databases.

Are your vectors routinely sequenced?

No, our vectors are not routinely sequenced. Quality control and release criteria utilize other methods.

How was the reference sequence for your vectors created?

Sequences provided for our vectors have been compiled from information in sequence databases, published sequences, and other sources.

What is the consensus Kozak sequence and what is the function of the Kozak sequence?

Eukaryotic (and specifically mammalian) mRNA contains sequence information that is important for efficient translation. However, this sequence, termed a Kozak sequence, is not a true ribosome binding site, but rather a translation initiation enhancer. The Kozak consensus sequence is ACCAUGG, where AUG is the initiation codon. A purine (A/G) in position -3 has a dominant effect; with a pyrimidine (C/T) in position -3, translation becomes more sensitive to changes in positions -1, -2, and +4. Expression levels can be reduced up to 95% when the -3 position is changed from a purine to pyrimidine. The +4 position has less influence on expression levels where approximately 50% reduction is seen. See the following references:

Kozak, M. (1986) Point mutations define a sequence flanking the AUG initiator codon that modulates translation by eukaryotic ribosomes. Cell 44, 283-292.
Kozak, M. (1987) At least six nucleotides preceding the AUG initiator codon enhance translation in mammalian cells. J. Mol. Biol. 196, 947-950.
Kozak, M. (1987) An analysis of 5´-noncoding sequences from 699 vertebrate messenger RNAs. Nucleic Acids Res. 15, 8125-8148.
Kozak, M. (1989) The scanning model for translation: An update. J. Cell Biol. 108, 229-241.
Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res. 18, 2828.

Note: The optimal Kozak sequence for Drosophila differs slightly, and yeast do not follow this rule at all. See the following references:

Romanos, M.A., Scorer, C.A., Clare, J.J. (1992) Foreign gene expression in yeast: a review. Yeast 8, 423-488.
Cavaneer, D.R. (1987) Comparison of the consensus sequence flanking translational start sites in Drosophila and vertebrates. Nucleic Acids Res. 15, 1353-1361.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.